Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87253311-87253568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
UniGene
Hs.476930
Gene Chromosome Location
Chr.3: 87227263-87255548 on Build GRCh38
Transcripts
NM_001244644.1 NM_014043.3 XM_011533576.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3597811 c.526G>A p.G176R Chr.3: 87253505-87253505 on Build GRCh38
1048781 c.485T>C p.I162T Chr.3: 87253464-87253464 on Build GRCh38
1048783 c.509T>G p.I170S Chr.3: 87253488-87253488 on Build GRCh38
399129 c.469G>A p.E157K Chr.3: 87253448-87253448 on Build GRCh38
5601240 c.512G>A p.G171E Chr.3: 87253491-87253491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758354665 Chr.3: 87253444-87253444 on Build GRCh38
rs746595208 Chr.3: 87253353-87253353 on Build GRCh38
rs757070412 Chr.3: 87253465-87253465 on Build GRCh38
rs977470994 Chr.3: 87253346-87253346 on Build GRCh38
rs375743465 Chr.3: 87253532-87253532 on Build GRCh38
rs199499269 Chr.3: 87253397-87253397 on Build GRCh38
rs372294128 Chr.3: 87253393-87253393 on Build GRCh38
rs368279409 Chr.3: 87253388-87253388 on Build GRCh38
rs541822657 Chr.3: 87253515-87253515 on Build GRCh38
rs942317217 Chr.3: 87253422-87253423 on Build GRCh38
rs749761645 Chr.3: 87253520-87253520 on Build GRCh38
rs374796686 Chr.3: 87253518-87253518 on Build GRCh38
rs1056391832 Chr.3: 87253399-87253399 on Build GRCh38
rs441639 Chr.3: 87253377-87253377 on Build GRCh38
rs767746876 Chr.3: 87253432-87253432 on Build GRCh38
rs776086385 Chr.3: 87253391-87253391 on Build GRCh38
rs756914093 Chr.3: 87253362-87253362 on Build GRCh38
rs760794461 Chr.3: 87253433-87253433 on Build GRCh38
rs63750653 Chr.3: 87253421-87253421 on Build GRCh38
rs63750944 Chr.3: 87253407-87253407 on Build GRCh38
rs774798983 Chr.3: 87253405-87253405 on Build GRCh38
rs750061631 Chr.3: 87253514-87253514 on Build GRCh38
rs780887155 Chr.3: 87253485-87253485 on Build GRCh38
rs765851600 Chr.3: 87253397-87253397 on Build GRCh38
rs772378083 Chr.3: 87253544-87253544 on Build GRCh38
rs35414442 Chr.3: 87253501-87253501 on Build GRCh38
rs751339317 Chr.3: 87253457-87253457 on Build GRCh38
rs780751677 Chr.3: 87253371-87253371 on Build GRCh38
rs63750355 Chr.3: 87253472-87253472 on Build GRCh38
rs370141207 Chr.3: 87253538-87253538 on Build GRCh38
rs375575660 Chr.3: 87253443-87253443 on Build GRCh38
rs764069353 Chr.3: 87253445-87253445 on Build GRCh38
rs749814652 Chr.3: 87253392-87253392 on Build GRCh38
rs1045230920 Chr.3: 87253505-87253505 on Build GRCh38
rs745382317 Chr.3: 87253373-87253373 on Build GRCh38
rs751018716 Chr.3: 87253534-87253534 on Build GRCh38

View Full Product Details