Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228102346-228102573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
UniGene
Hs.445952
Gene Chromosome Location
Chr.1: 228100727-228103462 on Build GRCh38
Transcripts
NM_024319.3 NR_130975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295953 c.404G>C p.R135P Chr.1: 228102353-228102353 on Build GRCh38
4028855 c.382G>A p.V128M Chr.1: 228102375-228102375 on Build GRCh38
3718402 c.374+2T>G p.? Chr.1: 228102476-228102476 on Build GRCh38
1601783 c.387C>T p.G129G Chr.1: 228102370-228102370 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs867593559 Chr.1: 228102538-228102538 on Build GRCh38
rs550471447 Chr.1: 228102471-228102471 on Build GRCh38
rs368673303 Chr.1: 228102500-228102500 on Build GRCh38
rs768292808 Chr.1: 228102370-228102370 on Build GRCh38
rs867326277 Chr.1: 228102482-228102482 on Build GRCh38
rs764220937 Chr.1: 228102419-228102419 on Build GRCh38
rs747586817 Chr.1: 228102393-228102393 on Build GRCh38
rs770016363 Chr.1: 228102404-228102404 on Build GRCh38
rs570632683 Chr.1: 228102491-228102491 on Build GRCh38
rs767426726 Chr.1: 228102530-228102530 on Build GRCh38
rs778552072 Chr.1: 228102375-228102375 on Build GRCh38
rs955445962 Chr.1: 228102513-228102513 on Build GRCh38
rs530647448 Chr.1: 228102426-228102426 on Build GRCh38
rs747390709 Chr.1: 228102516-228102518 on Build GRCh38
rs779654391 Chr.1: 228102439-228102439 on Build GRCh38
rs747640027 Chr.1: 228102459-228102459 on Build GRCh38
rs773212690 Chr.1: 228102539-228102539 on Build GRCh38
rs751604659 Chr.1: 228102420-228102420 on Build GRCh38
rs771534451 Chr.1: 228102398-228102398 on Build GRCh38
rs755939861 Chr.1: 228102430-228102430 on Build GRCh38
rs377443672 Chr.1: 228102522-228102522 on Build GRCh38
rs763032189 Chr.1: 228102416-228102416 on Build GRCh38
rs1042253104 Chr.1: 228102431-228102431 on Build GRCh38
rs1000703488 Chr.1: 228102474-228102474 on Build GRCh38
rs746296593 Chr.1: 228102484-228102484 on Build GRCh38
rs753584462 Chr.1: 228102440-228102440 on Build GRCh38
rs376871646 Chr.1: 228102413-228102413 on Build GRCh38
rs892140484 Chr.1: 228102412-228102412 on Build GRCh38
rs774504299 Chr.1: 228102520-228102520 on Build GRCh38
rs754673815 Chr.1: 228102443-228102443 on Build GRCh38
rs184854491 Chr.1: 228102512-228102512 on Build GRCh38
rs867187913 Chr.1: 228102521-228102521 on Build GRCh38
rs755468078 Chr.1: 228102538-228102539 on Build GRCh38
rs768840354 Chr.1: 228102518-228102518 on Build GRCh38
rs759969526 Chr.1: 228102401-228102401 on Build GRCh38
rs778607073 Chr.1: 228102456-228102456 on Build GRCh38
rs923204543 Chr.1: 228102417-228102417 on Build GRCh38
rs997002286 Chr.1: 228102473-228102473 on Build GRCh38
rs750285905 Chr.1: 228102423-228102423 on Build GRCh38
rs964177292 Chr.1: 228102432-228102432 on Build GRCh38
rs371201234 Chr.1: 228102555-228102555 on Build GRCh38
rs757934128 Chr.1: 228102466-228102466 on Build GRCh38

View Full Product Details