Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 96012-96279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB125
Entrez Gene ID
245938
Gene Name
defensin beta 125
Gene Aliases
DEFB-25
UniGene
Hs.380220
Gene Chromosome Location
Chr.20: 87672-96573 on Build GRCh38
Transcripts
NM_153325.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6533028 c.276A>C p.P92P Chr.20: 96222-96222 on Build GRCh38
3718594 c.113G>A p.R38Q Chr.20: 96059-96059 on Build GRCh38
5924134 c.84G>A p.W28* Chr.20: 96030-96030 on Build GRCh38
3549329 c.200G>A p.R67Q Chr.20: 96146-96146 on Build GRCh38
6094021 c.241G>T p.D81Y Chr.20: 96187-96187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751532285 Chr.20: 96075-96075 on Build GRCh38
rs960455369 Chr.20: 96127-96127 on Build GRCh38
rs532933005 Chr.20: 96081-96081 on Build GRCh38
rs370092526 Chr.20: 96062-96062 on Build GRCh38
rs748762744 Chr.20: 96095-96095 on Build GRCh38
rs200870238 Chr.20: 96149-96149 on Build GRCh38
rs773518041 Chr.20: 96121-96121 on Build GRCh38
rs771807843 Chr.20: 96143-96143 on Build GRCh38
rs754005070 Chr.20: 96142-96142 on Build GRCh38
rs750171266 Chr.20: 96162-96162 on Build GRCh38
rs148698629 Chr.20: 96125-96125 on Build GRCh38
rs565892940 Chr.20: 96103-96103 on Build GRCh38
rs778648489 Chr.20: 96112-96112 on Build GRCh38
rs768046229 Chr.20: 96096-96096 on Build GRCh38
rs867765515 Chr.20: 96219-96219 on Build GRCh38
rs772464690 Chr.20: 96115-96115 on Build GRCh38
rs116934569 Chr.20: 96059-96059 on Build GRCh38
rs369692500 Chr.20: 96243-96243 on Build GRCh38
rs774092161 Chr.20: 96069-96069 on Build GRCh38
rs762230159 Chr.20: 96050-96050 on Build GRCh38
rs375564330 Chr.20: 96197-96197 on Build GRCh38
rs778362608 Chr.20: 96179-96179 on Build GRCh38
rs767854746 Chr.20: 96051-96051 on Build GRCh38
rs202219607 Chr.20: 96085-96085 on Build GRCh38
rs769435585 Chr.20: 96094-96094 on Build GRCh38
rs151148014 Chr.20: 96145-96145 on Build GRCh38
rs750692266 Chr.20: 96052-96052 on Build GRCh38
rs969665579 Chr.20: 96240-96240 on Build GRCh38
rs373770106 Chr.20: 96083-96083 on Build GRCh38
rs773571509 Chr.20: 96229-96229 on Build GRCh38
rs145802690 Chr.20: 96048-96048 on Build GRCh38
rs762285182 Chr.20: 96108-96108 on Build GRCh38
rs890000950 Chr.20: 96078-96078 on Build GRCh38
rs558280500 Chr.20: 96245-96245 on Build GRCh38
rs766392612 Chr.20: 96058-96058 on Build GRCh38
rs760912688 Chr.20: 96056-96056 on Build GRCh38
rs139650748 Chr.20: 96155-96155 on Build GRCh38
rs748878197 Chr.20: 96174-96174 on Build GRCh38
rs745543814 Chr.20: 96093-96093 on Build GRCh38
rs757216800 Chr.20: 96146-96146 on Build GRCh38
rs768203979 Chr.20: 96177-96177 on Build GRCh38
rs781107020 Chr.20: 96092-96092 on Build GRCh38
rs755723306 Chr.20: 96163-96163 on Build GRCh38
rs993824024 Chr.20: 96098-96098 on Build GRCh38
rs748677322 Chr.20: 96183-96183 on Build GRCh38
rs761171088 Chr.20: 96232-96232 on Build GRCh38
rs201575332 Chr.20: 96049-96049 on Build GRCh38
rs779821283 Chr.20: 96169-96169 on Build GRCh38
rs147882888 Chr.20: 96166-96166 on Build GRCh38
rs41276376 Chr.20: 96130-96130 on Build GRCh38

View Full Product Details