Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93999061-93999334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1
Entrez Gene ID
10282
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
HBET1
UniGene
Hs.489132
Gene Chromosome Location
Chr.7: 93962770-94004382 on Build GRCh38
Transcripts
NM_001317739.1 NM_005868.5 NR_133908.1 NR_133909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093243 c.143C>A p.S48Y Chr.7: 93999171-93999171 on Build GRCh38
5679209 c.83G>T p.C28F Chr.7: 93999231-93999231 on Build GRCh38
4929071 c.21T>C p.G7G Chr.7: 93999293-93999293 on Build GRCh38
2864937 c.128T>C p.V43A Chr.7: 93999186-93999186 on Build GRCh38
5873410 c.83G>A p.C28Y Chr.7: 93999231-93999231 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758430860 Chr.7: 93999135-93999135 on Build GRCh38
rs560835651 Chr.7: 93999104-93999104 on Build GRCh38
rs558422753 Chr.7: 93999201-93999201 on Build GRCh38
rs199587849 Chr.7: 93999163-93999163 on Build GRCh38
rs943263868 Chr.7: 93999166-93999166 on Build GRCh38
rs11551305 Chr.7: 93999230-93999230 on Build GRCh38
rs749755613 Chr.7: 93999285-93999285 on Build GRCh38
rs369247771 Chr.7: 93999231-93999231 on Build GRCh38
rs372976829 Chr.7: 93999251-93999251 on Build GRCh38
rs941665519 Chr.7: 93999102-93999102 on Build GRCh38
rs749847667 Chr.7: 93999169-93999169 on Build GRCh38
rs756511960 Chr.7: 93999276-93999276 on Build GRCh38
rs897149498 Chr.7: 93999198-93999198 on Build GRCh38
rs369271500 Chr.7: 93999289-93999289 on Build GRCh38
rs752988697 Chr.7: 93999266-93999266 on Build GRCh38
rs778483908 Chr.7: 93999303-93999303 on Build GRCh38
rs747193412 Chr.7: 93999146-93999146 on Build GRCh38
rs778290433 Chr.7: 93999284-93999284 on Build GRCh38
rs773748027 Chr.7: 93999178-93999178 on Build GRCh38
rs1020730618 Chr.7: 93999207-93999207 on Build GRCh38
rs775325211 Chr.7: 93999204-93999204 on Build GRCh38
rs748498078 Chr.7: 93999160-93999160 on Build GRCh38
rs759213626 Chr.7: 93999190-93999190 on Build GRCh38
rs373071449 Chr.7: 93999306-93999307 on Build GRCh38
rs761749249 Chr.7: 93999237-93999237 on Build GRCh38
rs991741336 Chr.7: 93999089-93999089 on Build GRCh38
rs371303822 Chr.7: 93999123-93999123 on Build GRCh38
rs770502307 Chr.7: 93999174-93999174 on Build GRCh38
rs1051589774 Chr.7: 93999161-93999161 on Build GRCh38
rs781321258 Chr.7: 93999261-93999261 on Build GRCh38
rs374724468 Chr.7: 93999255-93999255 on Build GRCh38
rs566318017 Chr.7: 93999142-93999142 on Build GRCh38
rs778380204 Chr.7: 93999164-93999164 on Build GRCh38
rs1053670105 Chr.7: 93999172-93999172 on Build GRCh38
rs541754296 Chr.7: 93999180-93999180 on Build GRCh38
rs753708508 Chr.7: 93999236-93999236 on Build GRCh38
rs574628565 Chr.7: 93999097-93999097 on Build GRCh38
rs781612321 Chr.7: 93999154-93999154 on Build GRCh38
rs755210198 Chr.7: 93999153-93999153 on Build GRCh38
rs1011555940 Chr.7: 93999184-93999184 on Build GRCh38
rs751444235 Chr.7: 93999162-93999162 on Build GRCh38

View Full Product Details