Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7853232-7853491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTS2
Entrez Gene ID
10911
Gene Name
urotensin 2
Gene Aliases
PRO1068, U-II, UCN2, UII
UniGene
Hs.715862
Gene Chromosome Location
Chr.1: 7846345-7913615 on Build GRCh38
Transcripts
NM_021995.2 XM_011540537.2 XM_011540538.1 XM_017000116.1 XM_017000117.1 XM_017000118.1 XM_017000119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6819346 c.34G>A p.V12I Chr.1: 7853398-7853398 on Build GRCh38
3751401 c.47G>A p.R16Q Chr.1: 7853385-7853385 on Build GRCh38
7092060 c.30C>T p.L10L Chr.1: 7853402-7853402 on Build GRCh38
3751400 c.62C>T p.T21M Chr.1: 7853370-7853370 on Build GRCh38
4577578 c.12C>T p.N4N Chr.1: 7853420-7853420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764750565 Chr.1: 7853459-7853459 on Build GRCh38
rs201912093 Chr.1: 7853365-7853365 on Build GRCh38
rs768043417 Chr.1: 7853302-7853302 on Build GRCh38
rs370577824 Chr.1: 7853419-7853419 on Build GRCh38
rs762054604 Chr.1: 7853299-7853299 on Build GRCh38
rs780317300 Chr.1: 7853389-7853389 on Build GRCh38
rs769334569 Chr.1: 7853360-7853360 on Build GRCh38
rs774699346 Chr.1: 7853399-7853399 on Build GRCh38
rs746038236 Chr.1: 7853264-7853264 on Build GRCh38
rs778683875 Chr.1: 7853362-7853362 on Build GRCh38
rs899960240 Chr.1: 7853316-7853316 on Build GRCh38
rs1015537582 Chr.1: 7853378-7853378 on Build GRCh38
rs758943746 Chr.1: 7853438-7853438 on Build GRCh38
rs542568011 Chr.1: 7853373-7853373 on Build GRCh38
rs1028472068 Chr.1: 7853308-7853308 on Build GRCh38
rs749327800 Chr.1: 7853395-7853395 on Build GRCh38
rs147322701 Chr.1: 7853420-7853420 on Build GRCh38
rs199972403 Chr.1: 7853275-7853275 on Build GRCh38
rs762334000 Chr.1: 7853412-7853412 on Build GRCh38
rs766124573 Chr.1: 7853421-7853421 on Build GRCh38
rs774993383 Chr.1: 7853288-7853288 on Build GRCh38
rs556310976 Chr.1: 7853261-7853261 on Build GRCh38
rs377396587 Chr.1: 7853398-7853398 on Build GRCh38
rs763110511 Chr.1: 7853280-7853280 on Build GRCh38
rs371264091 Chr.1: 7853270-7853270 on Build GRCh38
rs752178514 Chr.1: 7853471-7853471 on Build GRCh38
rs776807829 Chr.1: 7853262-7853262 on Build GRCh38
rs753566289 Chr.1: 7853423-7853423 on Build GRCh38
rs138271613 Chr.1: 7853402-7853402 on Build GRCh38
rs764500360 Chr.1: 7853281-7853281 on Build GRCh38
rs13306061 Chr.1: 7853385-7853385 on Build GRCh38
rs997431759 Chr.1: 7853331-7853331 on Build GRCh38
rs228648 Chr.1: 7853370-7853370 on Build GRCh38
rs775484077 Chr.1: 7853278-7853278 on Build GRCh38
rs755789170 Chr.1: 7853343-7853343 on Build GRCh38
rs200948566 Chr.1: 7853386-7853386 on Build GRCh38
rs984791638 Chr.1: 7853350-7853350 on Build GRCh38
rs776071622 Chr.1: 7853304-7853305 on Build GRCh38
rs186448206 Chr.1: 7853346-7853346 on Build GRCh38
rs754582674 Chr.1: 7853358-7853358 on Build GRCh38
rs770550544 Chr.1: 7853271-7853274 on Build GRCh38
rs750842874 Chr.1: 7853330-7853330 on Build GRCh38

View Full Product Details