Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43363098-43363335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20
Entrez Gene ID
991
Gene Name
cell division cycle 20
Gene Aliases
CDC20A, bA276H19.3, p55CDC
UniGene
Hs.524947
Gene Chromosome Location
Chr.1: 43358955-43363203 on Build GRCh38
Transcripts
NM_001255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455596 c.1479C>T p.L493L Chr.1: 43363108-43363108 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552379875 Chr.1: 43363222-43363222 on Build GRCh38
rs770993686 Chr.1: 43363264-43363268 on Build GRCh38
rs952516727 Chr.1: 43363240-43363240 on Build GRCh38
rs773547140 Chr.1: 43363159-43363159 on Build GRCh38
rs371111819 Chr.1: 43363124-43363124 on Build GRCh38
rs1014705187 Chr.1: 43363200-43363200 on Build GRCh38
rs892551001 Chr.1: 43363170-43363170 on Build GRCh38
rs150211823 Chr.1: 43363162-43363162 on Build GRCh38
rs769139223 Chr.1: 43363152-43363152 on Build GRCh38
rs997196100 Chr.1: 43363236-43363236 on Build GRCh38
rs971164025 Chr.1: 43363203-43363203 on Build GRCh38
rs566224419 Chr.1: 43363228-43363228 on Build GRCh38
rs758666549 Chr.1: 43363134-43363134 on Build GRCh38
rs1024986508 Chr.1: 43363191-43363191 on Build GRCh38
rs1009498445 Chr.1: 43363189-43363189 on Build GRCh38
rs748200234 Chr.1: 43363155-43363155 on Build GRCh38
rs374399185 Chr.1: 43363167-43363167 on Build GRCh38
rs1002108476 Chr.1: 43363206-43363206 on Build GRCh38
rs747464002 Chr.1: 43363143-43363143 on Build GRCh38
rs535125219 Chr.1: 43363245-43363245 on Build GRCh38
rs759729497 Chr.1: 43363149-43363151 on Build GRCh38
rs1028627343 Chr.1: 43363291-43363291 on Build GRCh38
rs1034052296 Chr.1: 43363239-43363239 on Build GRCh38
rs767851301 Chr.1: 43363152-43363153 on Build GRCh38
rs965760857 Chr.1: 43363201-43363201 on Build GRCh38
rs548561233 Chr.1: 43363294-43363294 on Build GRCh38
rs78262841 Chr.1: 43363191-43363191 on Build GRCh38
rs780668517 Chr.1: 43363142-43363142 on Build GRCh38
rs908253320 Chr.1: 43363271-43363271 on Build GRCh38

View Full Product Details