Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74703358-74703603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
UniGene
Hs.432379
Gene Chromosome Location
Chr.11: 74696428-74731385 on Build GRCh38
Transcripts
NM_001278473.2 NM_001304390.1 NM_001304391.1 NM_001304415.1 NM_001304416.1 NM_001304417.1 NM_015424.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925558 c.821C>T p.P274L Chr.11: 74703430-74703430 on Build GRCh38
4672235 c.777C>T p.Y259Y Chr.11: 74703474-74703474 on Build GRCh38
5925559 c.820C>T p.P274S Chr.11: 74703431-74703431 on Build GRCh38
3666995 c.816C>A p.F272L Chr.11: 74703435-74703435 on Build GRCh38
4903411 c.835C>T p.L279L Chr.11: 74703416-74703416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146411227 Chr.11: 74703477-74703477 on Build GRCh38
rs779677559 Chr.11: 74703487-74703487 on Build GRCh38
rs200494377 Chr.11: 74703535-74703535 on Build GRCh38
rs375642407 Chr.11: 74703451-74703451 on Build GRCh38
rs759625489 Chr.11: 74703462-74703462 on Build GRCh38
rs201538393 Chr.11: 74703397-74703397 on Build GRCh38
rs575861558 Chr.11: 74703528-74703528 on Build GRCh38
rs761821968 Chr.11: 74703383-74703383 on Build GRCh38
rs559010088 Chr.11: 74703523-74703523 on Build GRCh38
rs200003845 Chr.11: 74703379-74703379 on Build GRCh38
rs776767128 Chr.11: 74703459-74703459 on Build GRCh38
rs749191826 Chr.11: 74703436-74703436 on Build GRCh38
rs141892720 Chr.11: 74703394-74703394 on Build GRCh38
rs780908509 Chr.11: 74703411-74703411 on Build GRCh38
rs753144296 Chr.11: 74703387-74703387 on Build GRCh38
rs573139000 Chr.11: 74703485-74703485 on Build GRCh38
rs763486343 Chr.11: 74703474-74703474 on Build GRCh38
rs754926668 Chr.11: 74703522-74703522 on Build GRCh38
rs769903702 Chr.11: 74703540-74703540 on Build GRCh38
rs146078923 Chr.11: 74703569-74703569 on Build GRCh38
rs780156644 Chr.11: 74703435-74703435 on Build GRCh38
rs375797871 Chr.11: 74703442-74703442 on Build GRCh38
rs879302787 Chr.11: 74703580-74703580 on Build GRCh38
rs139172308 Chr.11: 74703450-74703450 on Build GRCh38
rs567762819 Chr.11: 74703400-74703400 on Build GRCh38
rs767286688 Chr.11: 74703481-74703481 on Build GRCh38
rs553497214 Chr.11: 74703380-74703380 on Build GRCh38
rs542307881 Chr.11: 74703576-74703576 on Build GRCh38
rs61730072 Chr.11: 74703468-74703468 on Build GRCh38
rs571786707 Chr.11: 74703527-74703527 on Build GRCh38
rs756931331 Chr.11: 74703478-74703478 on Build GRCh38
rs749229986 Chr.11: 74703509-74703509 on Build GRCh38
rs146734849 Chr.11: 74703395-74703395 on Build GRCh38
rs755621615 Chr.11: 74703434-74703434 on Build GRCh38
rs776859425 Chr.11: 74703530-74703530 on Build GRCh38
rs374603983 Chr.11: 74703403-74703403 on Build GRCh38
rs765250083 Chr.11: 74703467-74703467 on Build GRCh38
rs745370924 Chr.11: 74703414-74703414 on Build GRCh38
rs181307995 Chr.11: 74703561-74703561 on Build GRCh38
rs201529169 Chr.11: 74703381-74703381 on Build GRCh38
rs539345267 Chr.11: 74703486-74703486 on Build GRCh38
rs573451761 Chr.11: 74703570-74703570 on Build GRCh38

View Full Product Details