Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96654604-96654849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAED1
Entrez Gene ID
195827
Gene Name
AhpC/TSA antioxidant enzyme domain containing 1
Gene Aliases
C9orf21
UniGene
Hs.44640
Gene Chromosome Location
Chr.9: 96641251-96655331 on Build GRCh38
Transcripts
NM_153698.1 XM_005251783.3 XM_005251784.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164156 c.242T>G p.I81S Chr.9: 96654724-96654724 on Build GRCh38
1464258 c.228G>T p.E76D Chr.9: 96654738-96654738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768762530 Chr.9: 96654674-96654674 on Build GRCh38
rs533799302 Chr.9: 96654646-96654646 on Build GRCh38
rs763910204 Chr.9: 96654759-96654759 on Build GRCh38
rs1010199410 Chr.9: 96654641-96654641 on Build GRCh38
rs773682968 Chr.9: 96654785-96654792 on Build GRCh38
rs566778299 Chr.9: 96654628-96654628 on Build GRCh38
rs767012567 Chr.9: 96654699-96654699 on Build GRCh38
rs780356139 Chr.9: 96654720-96654720 on Build GRCh38
rs561155813 Chr.9: 96654802-96654803 on Build GRCh38
rs73543002 Chr.9: 96654678-96654678 on Build GRCh38
rs540053823 Chr.9: 96654815-96654815 on Build GRCh38
rs145599965 Chr.9: 96654625-96654625 on Build GRCh38
rs919881941 Chr.9: 96654679-96654679 on Build GRCh38
rs766997408 Chr.9: 96654742-96654742 on Build GRCh38
rs868262906 Chr.9: 96654665-96654665 on Build GRCh38
rs941454215 Chr.9: 96654651-96654651 on Build GRCh38
rs902018580 Chr.9: 96654797-96654797 on Build GRCh38
rs773574655 Chr.9: 96654822-96654822 on Build GRCh38
rs985283545 Chr.9: 96654631-96654631 on Build GRCh38
rs1056721820 Chr.9: 96654721-96654721 on Build GRCh38
rs775041368 Chr.9: 96654716-96654716 on Build GRCh38
rs9886834 Chr.9: 96654718-96654718 on Build GRCh38
rs773449591 Chr.9: 96654688-96654688 on Build GRCh38
rs142695697 Chr.9: 96654754-96654754 on Build GRCh38
rs778789652 Chr.9: 96654775-96654775 on Build GRCh38
rs199744732 Chr.9: 96654663-96654663 on Build GRCh38
rs763512025 Chr.9: 96654694-96654694 on Build GRCh38
rs781144318 Chr.9: 96654788-96654788 on Build GRCh38
rs369721183 Chr.9: 96654792-96654792 on Build GRCh38
rs377563610 Chr.9: 96654825-96654825 on Build GRCh38
rs780061434 Chr.9: 96654671-96654671 on Build GRCh38
rs548581742 Chr.9: 96654659-96654659 on Build GRCh38
rs747068183 Chr.9: 96654673-96654673 on Build GRCh38
rs758524590 Chr.9: 96654824-96654824 on Build GRCh38
rs755727167 Chr.9: 96654753-96654753 on Build GRCh38
rs770133967 Chr.9: 96654687-96654687 on Build GRCh38
rs752340584 Chr.9: 96654749-96654749 on Build GRCh38
rs551263661 Chr.9: 96654627-96654627 on Build GRCh38
rs527298219 Chr.9: 96654719-96654719 on Build GRCh38
rs896771498 Chr.9: 96654736-96654736 on Build GRCh38
rs374010422 Chr.9: 96654796-96654796 on Build GRCh38
rs779277749 Chr.9: 96654682-96654682 on Build GRCh38
rs757093736 Chr.9: 96654771-96654771 on Build GRCh38
rs376957960 Chr.9: 96654700-96654700 on Build GRCh38
rs755072072 Chr.9: 96654784-96654784 on Build GRCh38

View Full Product Details