Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34493129-34493402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
UniGene
Hs.112667
Gene Chromosome Location
Chr.9: 34458752-34520989 on Build GRCh38
Transcripts
NM_001281428.1 NM_012144.3 XM_006716758.3 XM_011517846.1 XM_011517847.2 XM_011517848.1 XM_011517849.1 XM_011517850.2 XM_017014625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108464 c.784G>A p.A262T Chr.9: 34493296-34493296 on Build GRCh38
3926725 c.698C>T p.A233V Chr.9: 34493210-34493210 on Build GRCh38
1108463 c.753G>T p.K251N Chr.9: 34493265-34493265 on Build GRCh38
4558680 c.776G>A p.G259E Chr.9: 34493288-34493288 on Build GRCh38
4773462 c.737_739delAGA p.K247delK Chr.9: 34493249-34493251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754505627 Chr.9: 34493320-34493320 on Build GRCh38
rs775818668 Chr.9: 34493239-34493239 on Build GRCh38
rs199865010 Chr.9: 34493179-34493179 on Build GRCh38
rs755730248 Chr.9: 34493182-34493182 on Build GRCh38
rs377082537 Chr.9: 34493370-34493370 on Build GRCh38
rs763219522 Chr.9: 34493368-34493368 on Build GRCh38
rs779711895 Chr.9: 34493313-34493313 on Build GRCh38
rs757013140 Chr.9: 34493175-34493175 on Build GRCh38
rs1030019140 Chr.9: 34493308-34493308 on Build GRCh38
rs577672150 Chr.9: 34493204-34493204 on Build GRCh38
rs747512649 Chr.9: 34493327-34493327 on Build GRCh38
rs562408105 Chr.9: 34493253-34493253 on Build GRCh38
rs867966352 Chr.9: 34493270-34493270 on Build GRCh38
rs761802925 Chr.9: 34493283-34493283 on Build GRCh38
rs999905460 Chr.9: 34493232-34493232 on Build GRCh38
rs752805725 Chr.9: 34493164-34493164 on Build GRCh38
rs767337303 Chr.9: 34493293-34493293 on Build GRCh38
rs771491259 Chr.9: 34493210-34493210 on Build GRCh38
rs1057123340 Chr.9: 34493282-34493282 on Build GRCh38
rs556572825 Chr.9: 34493260-34493260 on Build GRCh38
rs199746889 Chr.9: 34493351-34493351 on Build GRCh38
rs764172924 Chr.9: 34493259-34493259 on Build GRCh38
rs976685507 Chr.9: 34493272-34493272 on Build GRCh38
rs748769889 Chr.9: 34493184-34493184 on Build GRCh38
rs200945732 Chr.9: 34493322-34493322 on Build GRCh38
rs765581497 Chr.9: 34493220-34493220 on Build GRCh38
rs1022431793 Chr.9: 34493205-34493205 on Build GRCh38
rs747538328 Chr.9: 34493209-34493209 on Build GRCh38
rs964084810 Chr.9: 34493267-34493267 on Build GRCh38
rs201119185 Chr.9: 34493218-34493218 on Build GRCh38
rs904246751 Chr.9: 34493229-34493229 on Build GRCh38
rs927359374 Chr.9: 34493267-34493267 on Build GRCh38
rs777107705 Chr.9: 34493213-34493213 on Build GRCh38
rs367663319 Chr.9: 34493340-34493340 on Build GRCh38
rs938687532 Chr.9: 34493269-34493269 on Build GRCh38
rs914390053 Chr.9: 34493214-34493214 on Build GRCh38
rs367872627 Chr.9: 34493336-34493336 on Build GRCh38
rs750194621 Chr.9: 34493299-34493299 on Build GRCh38
rs189617844 Chr.9: 34493376-34493376 on Build GRCh38
rs770220348 Chr.9: 34493357-34493357 on Build GRCh38
rs947504661 Chr.9: 34493244-34493244 on Build GRCh38
rs888628696 Chr.9: 34493367-34493367 on Build GRCh38
rs765451227 Chr.9: 34493155-34493155 on Build GRCh38
rs373703151 Chr.9: 34493275-34493275 on Build GRCh38
rs557628979 Chr.9: 34493190-34493190 on Build GRCh38
rs892615618 Chr.9: 34493173-34493173 on Build GRCh38
rs186333447 Chr.9: 34493183-34493183 on Build GRCh38
rs755853256 Chr.9: 34493303-34493303 on Build GRCh38
rs374387905 Chr.9: 34493317-34493317 on Build GRCh38
rs775584863 Chr.9: 34493359-34493359 on Build GRCh38
rs753174457 Chr.9: 34493378-34493378 on Build GRCh38
rs549279182 Chr.9: 34493176-34493176 on Build GRCh38
rs758459005 Chr.9: 34493167-34493167 on Build GRCh38
rs926629979 Chr.9: 34493377-34493377 on Build GRCh38

View Full Product Details