Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27628537-27628794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC121
Entrez Gene ID
79635
Gene Name
coiled-coil domain containing 121
Gene Aliases
-
UniGene
Hs.21081
Gene Chromosome Location
Chr.2: 27625638-27629031 on Build GRCh38
Transcripts
NM_001142683.2 NM_024584.4 XM_005264560.3 NM_001145047.1 NM_001145048.1 NM_001145049.1

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
UniGene
Hs.18259
Gene Chromosome Location
Chr.2: 27628648-27650846 on Build GRCh38
Transcripts
NM_001142683.2 NM_024584.4 XM_005264560.3 NM_001145047.1 NM_001145048.1 NM_001145049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592116 c.92G>A p.G31E Chr.2: 27628672-27628672 on Build GRCh38
1199806 c.152C>T p.P51L Chr.2: 27628612-27628612 on Build GRCh38
6267663 c.167C>A p.A56E Chr.2: 27628597-27628597 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769807338 Chr.2: 27628686-27628686 on Build GRCh38
rs548693625 Chr.2: 27628587-27628587 on Build GRCh38
rs372104931 Chr.2: 27628677-27628677 on Build GRCh38
rs61743551 Chr.2: 27628773-27628773 on Build GRCh38
rs568737551 Chr.2: 27628613-27628613 on Build GRCh38
rs780344224 Chr.2: 27628598-27628598 on Build GRCh38
rs779980800 Chr.2: 27628742-27628742 on Build GRCh38
rs781611356 Chr.2: 27628656-27628656 on Build GRCh38
rs60394995 Chr.2: 27628593-27628593 on Build GRCh38
rs1018795454 Chr.2: 27628766-27628766 on Build GRCh38
rs537460428 Chr.2: 27628728-27628728 on Build GRCh38
rs374842230 Chr.2: 27628752-27628752 on Build GRCh38
rs773459353 Chr.2: 27628701-27628701 on Build GRCh38
rs936116172 Chr.2: 27628564-27628564 on Build GRCh38
rs937621979 Chr.2: 27628655-27628655 on Build GRCh38
rs886304785 Chr.2: 27628760-27628760 on Build GRCh38
rs756932049 Chr.2: 27628595-27628595 on Build GRCh38
rs748241305 Chr.2: 27628679-27628679 on Build GRCh38
rs745744300 Chr.2: 27628661-27628661 on Build GRCh38
rs749474547 Chr.2: 27628759-27628759 on Build GRCh38
rs906131399 Chr.2: 27628634-27628634 on Build GRCh38
rs771293206 Chr.2: 27628761-27628761 on Build GRCh38
rs755442045 Chr.2: 27628632-27628632 on Build GRCh38
rs756026177 Chr.2: 27628690-27628690 on Build GRCh38
rs950267259 Chr.2: 27628607-27628607 on Build GRCh38
rs778999829 Chr.2: 27628659-27628661 on Build GRCh38
rs534109464 Chr.2: 27628559-27628559 on Build GRCh38
rs995035225 Chr.2: 27628749-27628749 on Build GRCh38

View Full Product Details