Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22447364-22447613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423823 c.500C>T p.P167L Chr.9: 22447565-22447565 on Build GRCh38
6856500 c.520delG p.G175fs*25 Chr.9: 22447585-22447585 on Build GRCh38
6357562 c.358G>C p.D120H Chr.9: 22447423-22447423 on Build GRCh38
227543 c.538G>A p.E180K Chr.9: 22447603-22447603 on Build GRCh38
3395768 c.488G>T p.G163V Chr.9: 22447553-22447553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752276044 Chr.9: 22447414-22447414 on Build GRCh38
rs749373349 Chr.9: 22447446-22447446 on Build GRCh38
rs766595649 Chr.9: 22447569-22447569 on Build GRCh38
rs34336274 Chr.9: 22447524-22447524 on Build GRCh38
rs770303277 Chr.9: 22447436-22447436 on Build GRCh38
rs750819763 Chr.9: 22447568-22447568 on Build GRCh38
rs531593966 Chr.9: 22447505-22447505 on Build GRCh38
rs753360659 Chr.9: 22447389-22447389 on Build GRCh38
rs757627293 Chr.9: 22447425-22447425 on Build GRCh38
rs752784700 Chr.9: 22447584-22447584 on Build GRCh38
rs758755308 Chr.9: 22447589-22447589 on Build GRCh38
rs764755749 Chr.9: 22447561-22447561 on Build GRCh38
rs746390595 Chr.9: 22447435-22447435 on Build GRCh38
rs779177309 Chr.9: 22447583-22447583 on Build GRCh38
rs777129344 Chr.9: 22447564-22447564 on Build GRCh38
rs776639443 Chr.9: 22447554-22447554 on Build GRCh38
rs915243023 Chr.9: 22447574-22447574 on Build GRCh38
rs915211979 Chr.9: 22447391-22447391 on Build GRCh38
rs369146134 Chr.9: 22447453-22447453 on Build GRCh38
rs771288515 Chr.9: 22447490-22447491 on Build GRCh38
rs770844580 Chr.9: 22447549-22447549 on Build GRCh38
rs759047301 Chr.9: 22447475-22447475 on Build GRCh38
rs546349473 Chr.9: 22447579-22447579 on Build GRCh38
rs535582130 Chr.9: 22447593-22447593 on Build GRCh38
rs182619176 Chr.9: 22447429-22447429 on Build GRCh38
rs756637667 Chr.9: 22447499-22447499 on Build GRCh38
rs981950952 Chr.9: 22447543-22447543 on Build GRCh38
rs759416915 Chr.9: 22447559-22447559 on Build GRCh38
rs749690923 Chr.9: 22447525-22447525 on Build GRCh38
rs760509051 Chr.9: 22447548-22447548 on Build GRCh38
rs529345917 Chr.9: 22447455-22447455 on Build GRCh38
rs896875470 Chr.9: 22447488-22447488 on Build GRCh38
rs746316567 Chr.9: 22447563-22447564 on Build GRCh38
rs570229256 Chr.9: 22447528-22447528 on Build GRCh38
rs1018566203 Chr.9: 22447556-22447556 on Build GRCh38
rs754640625 Chr.9: 22447407-22447407 on Build GRCh38
rs539265325 Chr.9: 22447564-22447564 on Build GRCh38
rs578231901 Chr.9: 22447398-22447398 on Build GRCh38
rs773022500 Chr.9: 22447463-22447463 on Build GRCh38
rs748414619 Chr.9: 22447532-22447532 on Build GRCh38
rs752365919 Chr.9: 22447478-22447478 on Build GRCh38
rs570387910 Chr.9: 22447557-22447557 on Build GRCh38
rs753272129 Chr.9: 22447477-22447477 on Build GRCh38
rs760179259 Chr.9: 22447392-22447392 on Build GRCh38
rs549110334 Chr.9: 22447468-22447468 on Build GRCh38
rs1050428620 Chr.9: 22447400-22447400 on Build GRCh38
rs923884921 Chr.9: 22447551-22447551 on Build GRCh38
rs759531750 Chr.9: 22447464-22447464 on Build GRCh38
rs764767867 Chr.9: 22447546-22447546 on Build GRCh38
rs755503848 Chr.9: 22447580-22447580 on Build GRCh38
rs772016586 Chr.9: 22447458-22447458 on Build GRCh38
rs772394134 Chr.9: 22447536-22447536 on Build GRCh38
rs374668199 Chr.9: 22447479-22447479 on Build GRCh38
rs779087748 Chr.9: 22447530-22447530 on Build GRCh38
rs750776463 Chr.9: 22447495-22447495 on Build GRCh38
rs200745233 Chr.9: 22447591-22447591 on Build GRCh38
rs560582163 Chr.9: 22447445-22447445 on Build GRCh38
rs1037059968 Chr.9: 22447454-22447454 on Build GRCh38

View Full Product Details