Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47603528-47603781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORIN
Entrez Gene ID
10699
Gene Name
corin, serine peptidase
Gene Aliases
ATC2, CRN, Lrp4, PEE5, TMPRSS10
UniGene
Hs.518618
Gene Chromosome Location
Chr.4: 47593998-47838106 on Build GRCh38
Transcripts
NM_001278585.1 NM_006587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6529221 c.2609T>G p.M870R Chr.4: 47603600-47603600 on Build GRCh38
7107120 c.2600C>T p.S867L Chr.4: 47603609-47603609 on Build GRCh38
3604190 c.2562G>A p.W854* Chr.4: 47603647-47603647 on Build GRCh38
3767892 c.2630C>T p.T877I Chr.4: 47603579-47603579 on Build GRCh38
1055730 c.2572C>T p.L858F Chr.4: 47603637-47603637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769858414 Chr.4: 47603707-47603707 on Build GRCh38
rs575717887 Chr.4: 47603613-47603613 on Build GRCh38
rs757904807 Chr.4: 47603687-47603687 on Build GRCh38
rs750194881 Chr.4: 47603562-47603562 on Build GRCh38
rs754167455 Chr.4: 47603666-47603668 on Build GRCh38
rs763146479 Chr.4: 47603628-47603628 on Build GRCh38
rs778244051 Chr.4: 47603692-47603692 on Build GRCh38
rs759722824 Chr.4: 47603622-47603622 on Build GRCh38
rs554132356 Chr.4: 47603550-47603550 on Build GRCh38
rs554963000 Chr.4: 47603664-47603664 on Build GRCh38
rs747303237 Chr.4: 47603609-47603609 on Build GRCh38
rs571162211 Chr.4: 47603592-47603592 on Build GRCh38
rs763119993 Chr.4: 47603718-47603718 on Build GRCh38
rs776895833 Chr.4: 47603616-47603616 on Build GRCh38
rs747569563 Chr.4: 47603696-47603696 on Build GRCh38
rs764585758 Chr.4: 47603616-47603617 on Build GRCh38
rs748953669 Chr.4: 47603579-47603579 on Build GRCh38
rs369384029 Chr.4: 47603557-47603557 on Build GRCh38
rs150816024 Chr.4: 47603552-47603552 on Build GRCh38
rs566304016 Chr.4: 47603553-47603553 on Build GRCh38
rs139246524 Chr.4: 47603581-47603581 on Build GRCh38
rs747604102 Chr.4: 47603686-47603686 on Build GRCh38
rs539641999 Chr.4: 47603569-47603569 on Build GRCh38
rs775669616 Chr.4: 47603625-47603625 on Build GRCh38
rs201830761 Chr.4: 47603588-47603588 on Build GRCh38
rs17572227 Chr.4: 47603717-47603717 on Build GRCh38
rs753499811 Chr.4: 47603673-47603673 on Build GRCh38
rs754550580 Chr.4: 47603681-47603681 on Build GRCh38
rs781309311 Chr.4: 47603561-47603561 on Build GRCh38
rs199894638 Chr.4: 47603559-47603559 on Build GRCh38
rs778329964 Chr.4: 47603585-47603585 on Build GRCh38
rs773726440 Chr.4: 47603604-47603604 on Build GRCh38
rs776898733 Chr.4: 47603702-47603702 on Build GRCh38
rs750452719 Chr.4: 47603631-47603631 on Build GRCh38
rs533197792 Chr.4: 47603753-47603753 on Build GRCh38
rs772523018 Chr.4: 47603600-47603600 on Build GRCh38
rs144619717 Chr.4: 47603746-47603746 on Build GRCh38
rs771302790 Chr.4: 47603697-47603697 on Build GRCh38
rs762352651 Chr.4: 47603682-47603684 on Build GRCh38
rs61759671 Chr.4: 47603736-47603736 on Build GRCh38
rs201935596 Chr.4: 47603624-47603624 on Build GRCh38
rs917279482 Chr.4: 47603571-47603571 on Build GRCh38
rs759097162 Chr.4: 47603561-47603562 on Build GRCh38
rs778562681 Chr.4: 47603683-47603683 on Build GRCh38
rs779753769 Chr.4: 47603574-47603574 on Build GRCh38
rs370637821 Chr.4: 47603705-47603705 on Build GRCh38
rs921026694 Chr.4: 47603703-47603703 on Build GRCh38
rs376924333 Chr.4: 47603652-47603652 on Build GRCh38

View Full Product Details