Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 644441-644692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEAF1
Entrez Gene ID
10522
Gene Name
DEAF1, transcription factor
Gene Aliases
MRD24, NUDR, SPN, ZMYND5
UniGene
Hs.243994
Gene Chromosome Location
Chr.11: 644220-695754 on Build GRCh38
Transcripts
NM_001293634.1 NM_021008.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7364041 c.1415C>G p.T472S Chr.11: 644608-644608 on Build GRCh38
4436348 c.1693G>A p.V565M Chr.11: 644555-644555 on Build GRCh38
6366265 c.1386G>T p.Q462H Chr.11: 644637-644637 on Build GRCh38
4879751 c.1679T>G p.M560R Chr.11: 644569-644569 on Build GRCh38
930169 c.1654G>A p.E552K Chr.11: 644594-644594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955197890 Chr.11: 644518-644518 on Build GRCh38
rs967974359 Chr.11: 644511-644511 on Build GRCh38
rs1021611482 Chr.11: 644622-644622 on Build GRCh38
rs748500387 Chr.11: 644621-644621 on Build GRCh38
rs1021664217 Chr.11: 644466-644466 on Build GRCh38
rs956246974 Chr.11: 644473-644473 on Build GRCh38
rs374342913 Chr.11: 644541-644541 on Build GRCh38
rs749882867 Chr.11: 644515-644515 on Build GRCh38
rs749741145 Chr.11: 644554-644554 on Build GRCh38
rs1044628242 Chr.11: 644543-644543 on Build GRCh38
rs762781481 Chr.11: 644649-644649 on Build GRCh38
rs747241811 Chr.11: 644530-644530 on Build GRCh38
rs1021561019 Chr.11: 644481-644481 on Build GRCh38
rs990136940 Chr.11: 644478-644478 on Build GRCh38
rs968320000 Chr.11: 644468-644468 on Build GRCh38
rs992255306 Chr.11: 644494-644494 on Build GRCh38
rs754762861 Chr.11: 644526-644526 on Build GRCh38
rs540841483 Chr.11: 644540-644540 on Build GRCh38
rs779454263 Chr.11: 644620-644620 on Build GRCh38
rs759498621 Chr.11: 644639-644639 on Build GRCh38
rs1031731821 Chr.11: 644465-644465 on Build GRCh38
rs148729216 Chr.11: 644561-644561 on Build GRCh38
rs757401613 Chr.11: 644536-644536 on Build GRCh38
rs142320326 Chr.11: 644595-644595 on Build GRCh38
rs996723909 Chr.11: 644630-644630 on Build GRCh38
rs767702509 Chr.11: 644652-644652 on Build GRCh38
rs1003371972 Chr.11: 644460-644460 on Build GRCh38
rs199717663 Chr.11: 644665-644665 on Build GRCh38
rs770483499 Chr.11: 644633-644633 on Build GRCh38
rs760200310 Chr.11: 644573-644573 on Build GRCh38
rs975291534 Chr.11: 644512-644512 on Build GRCh38
rs201859545 Chr.11: 644673-644673 on Build GRCh38
rs948983635 Chr.11: 644542-644542 on Build GRCh38
rs771661969 Chr.11: 644627-644627 on Build GRCh38
rs780211842 Chr.11: 644561-644561 on Build GRCh38
rs201424932 Chr.11: 644529-644529 on Build GRCh38
rs766292709 Chr.11: 644574-644574 on Build GRCh38
rs980036538 Chr.11: 644479-644479 on Build GRCh38
rs79183202 Chr.11: 644568-644568 on Build GRCh38
rs766952063 Chr.11: 644514-644514 on Build GRCh38
rs544459219 Chr.11: 644489-644489 on Build GRCh38
rs774798427 Chr.11: 644556-644556 on Build GRCh38
rs765017164 Chr.11: 644594-644594 on Build GRCh38
rs368188987 Chr.11: 644597-644597 on Build GRCh38
rs145256931 Chr.11: 644628-644628 on Build GRCh38
rs574660164 Chr.11: 644537-644537 on Build GRCh38
rs532671271 Chr.11: 644585-644585 on Build GRCh38
rs766316048 Chr.11: 644517-644517 on Build GRCh38
rs1033832762 Chr.11: 644488-644488 on Build GRCh38
rs776049891 Chr.11: 644637-644637 on Build GRCh38
rs111377617 Chr.11: 644565-644565 on Build GRCh38
rs757439651 Chr.11: 644603-644603 on Build GRCh38
rs1053683350 Chr.11: 644588-644588 on Build GRCh38
rs575533385 Chr.11: 644463-644463 on Build GRCh38
rs765107306 Chr.11: 644640-644640 on Build GRCh38
rs111947578 Chr.11: 644608-644608 on Build GRCh38
rs756513735 Chr.11: 644504-644505 on Build GRCh38
rs749827262 Chr.11: 644609-644609 on Build GRCh38
rs760160645 Chr.11: 644632-644632 on Build GRCh38
rs34114147 Chr.11: 644614-644614 on Build GRCh38
rs769130155 Chr.11: 644555-644555 on Build GRCh38
rs200505316 Chr.11: 644607-644607 on Build GRCh38
rs868832432 Chr.11: 644642-644651 on Build GRCh38
rs776038399 Chr.11: 644546-644546 on Build GRCh38
rs889636270 Chr.11: 644535-644535 on Build GRCh38

View Full Product Details