Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55162780-55163027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613880 c.1619_1621delATA p.N540delN Chr.14: 55163003-55163005 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554722961 Chr.14: 55162882-55162882 on Build GRCh38
rs755064912 Chr.14: 55162940-55162940 on Build GRCh38
rs762152301 Chr.14: 55162920-55162920 on Build GRCh38
rs781467096 Chr.14: 55162944-55162944 on Build GRCh38
rs755901847 Chr.14: 55162977-55162977 on Build GRCh38
rs756159520 Chr.14: 55162963-55162964 on Build GRCh38
rs1015678274 Chr.14: 55162923-55162923 on Build GRCh38
rs751629955 Chr.14: 55162935-55162935 on Build GRCh38
rs1027542533 Chr.14: 55162919-55162920 on Build GRCh38
rs778645013 Chr.14: 55162838-55162838 on Build GRCh38
rs763474961 Chr.14: 55162931-55162931 on Build GRCh38
rs994917481 Chr.14: 55162916-55162916 on Build GRCh38
rs766885632 Chr.14: 55162933-55162933 on Build GRCh38
rs897449147 Chr.14: 55162913-55162913 on Build GRCh38
rs752014944 Chr.14: 55162933-55162934 on Build GRCh38
rs536325712 Chr.14: 55162865-55162865 on Build GRCh38
rs866461734 Chr.14: 55162979-55162979 on Build GRCh38
rs867619439 Chr.14: 55162951-55162951 on Build GRCh38
rs766331356 Chr.14: 55162933-55162933 on Build GRCh38
rs752806634 Chr.14: 55162953-55162954 on Build GRCh38
rs1014046501 Chr.14: 55162876-55162878 on Build GRCh38
rs966098260 Chr.14: 55162933-55162937 on Build GRCh38
rs1014594599 Chr.14: 55162906-55162906 on Build GRCh38
rs750872286 Chr.14: 55162927-55162927 on Build GRCh38
rs753038944 Chr.14: 55162946-55162946 on Build GRCh38
rs988147506 Chr.14: 55162936-55162936 on Build GRCh38
rs765590172 Chr.14: 55162925-55162925 on Build GRCh38
rs895554202 Chr.14: 55162817-55162817 on Build GRCh38
rs759769063 Chr.14: 55162932-55162933 on Build GRCh38
rs913996180 Chr.14: 55162943-55162943 on Build GRCh38

View Full Product Details