Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32325077-32325302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245259 c.346A>T p.S116C Chr.13: 32325105-32325105 on Build GRCh38
5638347 c.425+1G>C p.? Chr.13: 32325185-32325185 on Build GRCh38
3955533 c.346A>C p.S116R Chr.13: 32325105-32325105 on Build GRCh38
469350 c.374A>G p.D125G Chr.13: 32325133-32325133 on Build GRCh38
7400281 c.355A>G p.T119A Chr.13: 32325114-32325114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80358621 Chr.13: 32325138-32325138 on Build GRCh38
rs886040488 Chr.13: 32325127-32325131 on Build GRCh38
rs944205640 Chr.13: 32325136-32325136 on Build GRCh38
rs752207703 Chr.13: 32325153-32325153 on Build GRCh38
rs80358603 Chr.13: 32325112-32325112 on Build GRCh38
rs587782590 Chr.13: 32325185-32325185 on Build GRCh38
rs397507713 Chr.13: 32325184-32325184 on Build GRCh38
rs876658305 Chr.13: 32325126-32325126 on Build GRCh38
rs80359427 Chr.13: 32325169-32325169 on Build GRCh38
rs397507680 Chr.13: 32325123-32325123 on Build GRCh38
rs80359425 Chr.13: 32325166-32325166 on Build GRCh38
rs876660310 Chr.13: 32325158-32325158 on Build GRCh38
rs886038050 Chr.13: 32325178-32325178 on Build GRCh38
rs397507324 Chr.13: 32325172-32325172 on Build GRCh38
rs879255321 Chr.13: 32325137-32325138 on Build GRCh38
rs397507681 Chr.13: 32325129-32325129 on Build GRCh38
rs786201593 Chr.13: 32325137-32325137 on Build GRCh38
rs200065709 Chr.13: 32325217-32325217 on Build GRCh38
rs766194968 Chr.13: 32325109-32325109 on Build GRCh38
rs201865785 Chr.13: 32325220-32325220 on Build GRCh38
rs1048366129 Chr.13: 32325213-32325213 on Build GRCh38
rs876661045 Chr.13: 32325186-32325186 on Build GRCh38
rs932657676 Chr.13: 32325187-32325187 on Build GRCh38
rs571823764 Chr.13: 32325156-32325156 on Build GRCh38
rs80358660 Chr.13: 32325175-32325175 on Build GRCh38
rs397507320 Chr.13: 32325155-32325155 on Build GRCh38
rs397507314 Chr.13: 32325129-32325129 on Build GRCh38
rs886038049 Chr.13: 32325150-32325150 on Build GRCh38
rs80358627 Chr.13: 32325142-32325142 on Build GRCh38
rs876658850 Chr.13: 32325172-32325176 on Build GRCh38
rs483353080 Chr.13: 32325169-32325170 on Build GRCh38
rs886040490 Chr.13: 32325129-32325133 on Build GRCh38
rs756310422 Chr.13: 32325236-32325236 on Build GRCh38
rs757612558 Chr.13: 32325188-32325188 on Build GRCh38
rs375125172 Chr.13: 32325111-32325111 on Build GRCh38
rs80358662 Chr.13: 32325177-32325177 on Build GRCh38
rs397507318 Chr.13: 32325148-32325148 on Build GRCh38
rs786202707 Chr.13: 32325140-32325140 on Build GRCh38
rs80358616 Chr.13: 32325134-32325134 on Build GRCh38
rs967234688 Chr.13: 32325180-32325180 on Build GRCh38
rs11571610 Chr.13: 32325251-32325251 on Build GRCh38

View Full Product Details