Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44624496-44624769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
UniGene
Hs.596000
Gene Chromosome Location
Chr.4: 44619578-44656868 on Build GRCh38
Transcripts
NM_182592.2 XM_011513679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447887 c.542T>A p.I181N Chr.4: 44624739-44624739 on Build GRCh38
5694291 c.533T>A p.M178K Chr.4: 44624748-44624748 on Build GRCh38
4504577 c.669C>T p.F223F Chr.4: 44624612-44624612 on Build GRCh38
6567403 c.669C>G p.F223L Chr.4: 44624612-44624612 on Build GRCh38
447885 c.643G>A p.V215I Chr.4: 44624638-44624638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751659018 Chr.4: 44624685-44624685 on Build GRCh38
rs901642298 Chr.4: 44624688-44624688 on Build GRCh38
rs554436483 Chr.4: 44624728-44624728 on Build GRCh38
rs776199494 Chr.4: 44624671-44624671 on Build GRCh38
rs756048138 Chr.4: 44624687-44624687 on Build GRCh38
rs143481397 Chr.4: 44624658-44624658 on Build GRCh38
rs976768544 Chr.4: 44624567-44624567 on Build GRCh38
rs771085825 Chr.4: 44624546-44624546 on Build GRCh38
rs370317024 Chr.4: 44624714-44624714 on Build GRCh38
rs1028815700 Chr.4: 44624707-44624707 on Build GRCh38
rs370783649 Chr.4: 44624666-44624666 on Build GRCh38
rs759911925 Chr.4: 44624678-44624678 on Build GRCh38
rs756066863 Chr.4: 44624649-44624649 on Build GRCh38
rs768401055 Chr.4: 44624657-44624657 on Build GRCh38
rs758504308 Chr.4: 44624625-44624625 on Build GRCh38
rs186215680 Chr.4: 44624651-44624651 on Build GRCh38
rs867312237 Chr.4: 44624718-44624718 on Build GRCh38
rs746546018 Chr.4: 44624725-44624725 on Build GRCh38
rs746163313 Chr.4: 44624730-44624730 on Build GRCh38
rs765403472 Chr.4: 44624679-44624679 on Build GRCh38
rs775888663 Chr.4: 44624742-44624742 on Build GRCh38
rs748937746 Chr.4: 44624552-44624552 on Build GRCh38
rs377377967 Chr.4: 44624662-44624662 on Build GRCh38
rs766276259 Chr.4: 44624693-44624693 on Build GRCh38
rs376027917 Chr.4: 44624604-44624604 on Build GRCh38
rs964696896 Chr.4: 44624585-44624585 on Build GRCh38
rs1043766788 Chr.4: 44624663-44624663 on Build GRCh38
rs60959366 Chr.4: 44624624-44624624 on Build GRCh38
rs747819263 Chr.4: 44624706-44624706 on Build GRCh38
rs559992068 Chr.4: 44624551-44624551 on Build GRCh38
rs754219707 Chr.4: 44624614-44624614 on Build GRCh38
rs763233871 Chr.4: 44624744-44624744 on Build GRCh38
rs371547157 Chr.4: 44624665-44624665 on Build GRCh38
rs749086893 Chr.4: 44624706-44624715 on Build GRCh38
rs974813374 Chr.4: 44624593-44624593 on Build GRCh38
rs80199883 Chr.4: 44624621-44624621 on Build GRCh38
rs376874064 Chr.4: 44624710-44624710 on Build GRCh38
rs759719745 Chr.4: 44624615-44624615 on Build GRCh38
rs768490878 Chr.4: 44624568-44624568 on Build GRCh38
rs936963213 Chr.4: 44624699-44624699 on Build GRCh38
rs772413823 Chr.4: 44624591-44624591 on Build GRCh38
rs375672898 Chr.4: 44624684-44624684 on Build GRCh38
rs750385797 Chr.4: 44624633-44624633 on Build GRCh38
rs375792072 Chr.4: 44624680-44624680 on Build GRCh38
rs536043627 Chr.4: 44624672-44624672 on Build GRCh38
rs780923586 Chr.4: 44624632-44624632 on Build GRCh38
rs754855720 Chr.4: 44624702-44624702 on Build GRCh38
rs994372331 Chr.4: 44624521-44624521 on Build GRCh38
rs778644184 Chr.4: 44624704-44624704 on Build GRCh38
rs773803678 Chr.4: 44624592-44624592 on Build GRCh38
rs749321446 Chr.4: 44624529-44624529 on Build GRCh38
rs760884979 Chr.4: 44624595-44624595 on Build GRCh38
rs115065558 Chr.4: 44624569-44624569 on Build GRCh38
rs762961360 Chr.4: 44624681-44624681 on Build GRCh38
rs753643088 Chr.4: 44624695-44624695 on Build GRCh38
rs372385431 Chr.4: 44624582-44624582 on Build GRCh38
rs770155368 Chr.4: 44624732-44624732 on Build GRCh38
rs909915100 Chr.4: 44624606-44624606 on Build GRCh38
rs1037307165 Chr.4: 44624620-44624620 on Build GRCh38
rs776955364 Chr.4: 44624674-44624674 on Build GRCh38

View Full Product Details