Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194341537-194341764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN2
Entrez Gene ID
1370
Gene Name
carboxypeptidase N subunit 2
Gene Aliases
ACBP
UniGene
Hs.528368
Gene Chromosome Location
Chr.3: 194339765-194351387 on Build GRCh38
Transcripts
NM_001080513.3 NM_001291988.1 XM_005269280.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5443804 c.1007T>A p.L336Q Chr.3: 194341696-194341696 on Build GRCh38
3591511 c.950C>T p.S317F Chr.3: 194341753-194341753 on Build GRCh38
3591510 c.986C>T p.P329L Chr.3: 194341717-194341717 on Build GRCh38
3591509 c.1066C>T p.L356F Chr.3: 194341637-194341637 on Build GRCh38
5398900 c.951C>T p.S317S Chr.3: 194341752-194341752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759600139 Chr.3: 194341603-194341603 on Build GRCh38
rs757568706 Chr.3: 194341642-194341642 on Build GRCh38
rs768640281 Chr.3: 194341669-194341669 on Build GRCh38
rs1049111194 Chr.3: 194341661-194341661 on Build GRCh38
rs746931561 Chr.3: 194341659-194341659 on Build GRCh38
rs775664002 Chr.3: 194341659-194341661 on Build GRCh38
rs911605161 Chr.3: 194341601-194341601 on Build GRCh38
rs775761428 Chr.3: 194341687-194341687 on Build GRCh38
rs752500940 Chr.3: 194341681-194341681 on Build GRCh38
rs116418749 Chr.3: 194341573-194341573 on Build GRCh38
rs148901764 Chr.3: 194341694-194341694 on Build GRCh38
rs778571990 Chr.3: 194341643-194341643 on Build GRCh38
rs756725736 Chr.3: 194341613-194341613 on Build GRCh38
rs199513160 Chr.3: 194341688-194341688 on Build GRCh38
rs979628492 Chr.3: 194341668-194341668 on Build GRCh38
rs1046465778 Chr.3: 194341680-194341680 on Build GRCh38
rs146800212 Chr.3: 194341650-194341650 on Build GRCh38
rs920257407 Chr.3: 194341654-194341654 on Build GRCh38
rs139721536 Chr.3: 194341721-194341721 on Build GRCh38
rs758059138 Chr.3: 194341684-194341684 on Build GRCh38
rs767723784 Chr.3: 194341605-194341605 on Build GRCh38
rs375711355 Chr.3: 194341651-194341651 on Build GRCh38
rs200698533 Chr.3: 194341640-194341640 on Build GRCh38
rs779582769 Chr.3: 194341658-194341658 on Build GRCh38
rs377624573 Chr.3: 194341676-194341676 on Build GRCh38
rs754286406 Chr.3: 194341620-194341620 on Build GRCh38
rs765395055 Chr.3: 194341653-194341653 on Build GRCh38
rs759704781 Chr.3: 194341683-194341683 on Build GRCh38
rs143546741 Chr.3: 194341639-194341639 on Build GRCh38
rs141301266 Chr.3: 194341572-194341572 on Build GRCh38
rs867272722 Chr.3: 194341712-194341712 on Build GRCh38
rs776463067 Chr.3: 194341574-194341574 on Build GRCh38
rs763075292 Chr.3: 194341583-194341583 on Build GRCh38
rs541517902 Chr.3: 194341726-194341726 on Build GRCh38
rs144778341 Chr.3: 194341560-194341560 on Build GRCh38
rs186606263 Chr.3: 194341618-194341618 on Build GRCh38
rs1002235038 Chr.3: 194341715-194341715 on Build GRCh38
rs370583151 Chr.3: 194341731-194341731 on Build GRCh38
rs747907608 Chr.3: 194341678-194341678 on Build GRCh38
rs775357382 Chr.3: 194341571-194341571 on Build GRCh38
rs764763182 Chr.3: 194341707-194341707 on Build GRCh38
rs774580275 Chr.3: 194341682-194341682 on Build GRCh38
rs766694115 Chr.3: 194341597-194341597 on Build GRCh38
rs753214592 Chr.3: 194341608-194341608 on Build GRCh38

View Full Product Details