Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83652672-83652941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN1
Entrez Gene ID
1404
Gene Name
hyaluronan and proteoglycan link protein 1
Gene Aliases
CRT1, CRTL1
UniGene
Hs.2799
Gene Chromosome Location
Chr.5: 83638198-83721210 on Build GRCh38
Transcripts
NM_001884.3 XM_011543168.2 XM_017009051.1 XM_017009052.1 XM_017009053.1 XM_017009054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618438 c.231C>T p.I77I Chr.5: 83652694-83652694 on Build GRCh38
4956272 c.111C>T p.G37G Chr.5: 83652814-83652814 on Build GRCh38
1070975 c.188T>G p.F63C Chr.5: 83652737-83652737 on Build GRCh38
3618437 c.237T>C p.I79I Chr.5: 83652688-83652688 on Build GRCh38
4479376 c.231C>A p.I77I Chr.5: 83652694-83652694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756746376 Chr.5: 83652708-83652708 on Build GRCh38
rs542489423 Chr.5: 83652811-83652811 on Build GRCh38
rs185835624 Chr.5: 83652844-83652844 on Build GRCh38
rs531265982 Chr.5: 83652894-83652894 on Build GRCh38
rs755758988 Chr.5: 83652728-83652728 on Build GRCh38
rs775525333 Chr.5: 83652840-83652840 on Build GRCh38
rs766950407 Chr.5: 83652719-83652719 on Build GRCh38
rs752039970 Chr.5: 83652700-83652700 on Build GRCh38
rs770973771 Chr.5: 83652767-83652767 on Build GRCh38
rs779740510 Chr.5: 83652729-83652729 on Build GRCh38
rs758251315 Chr.5: 83652855-83652855 on Build GRCh38
rs530272123 Chr.5: 83652797-83652797 on Build GRCh38
rs950672104 Chr.5: 83652804-83652804 on Build GRCh38
rs547235109 Chr.5: 83652795-83652795 on Build GRCh38
rs753629809 Chr.5: 83652859-83652862 on Build GRCh38
rs775898414 Chr.5: 83652782-83652782 on Build GRCh38
rs991351621 Chr.5: 83652859-83652859 on Build GRCh38
rs765976542 Chr.5: 83652832-83652832 on Build GRCh38
rs754759453 Chr.5: 83652745-83652745 on Build GRCh38
rs753508982 Chr.5: 83652838-83652838 on Build GRCh38
rs559408235 Chr.5: 83652777-83652777 on Build GRCh38
rs144483408 Chr.5: 83652769-83652769 on Build GRCh38
rs755660679 Chr.5: 83652831-83652831 on Build GRCh38
rs576797253 Chr.5: 83652758-83652758 on Build GRCh38
rs982110777 Chr.5: 83652815-83652815 on Build GRCh38
rs962883787 Chr.5: 83652761-83652761 on Build GRCh38
rs768137611 Chr.5: 83652817-83652817 on Build GRCh38
rs911415933 Chr.5: 83652808-83652808 on Build GRCh38
rs763374644 Chr.5: 83652793-83652793 on Build GRCh38
rs180793962 Chr.5: 83652723-83652723 on Build GRCh38
rs768968964 Chr.5: 83652851-83652851 on Build GRCh38
rs749922283 Chr.5: 83652830-83652830 on Build GRCh38
rs762232801 Chr.5: 83652813-83652813 on Build GRCh38
rs552522157 Chr.5: 83652897-83652897 on Build GRCh38
rs778859881 Chr.5: 83652751-83652751 on Build GRCh38
rs748039286 Chr.5: 83652853-83652853 on Build GRCh38
rs749051250 Chr.5: 83652731-83652731 on Build GRCh38
rs199764569 Chr.5: 83652709-83652709 on Build GRCh38
rs770115705 Chr.5: 83652780-83652780 on Build GRCh38
rs760495727 Chr.5: 83652839-83652840 on Build GRCh38
rs774866703 Chr.5: 83652809-83652809 on Build GRCh38
rs763996139 Chr.5: 83652840-83652842 on Build GRCh38
rs777902946 Chr.5: 83652864-83652864 on Build GRCh38
rs748048150 Chr.5: 83652752-83652752 on Build GRCh38

View Full Product Details