Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136362317-136362576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7
Entrez Gene ID
9053
Gene Name
microtubule associated protein 7
Gene Aliases
E-MAP-115, EMAP115
UniGene
Hs.486548
Gene Chromosome Location
Chr.6: 136342281-136551015 on Build GRCh38
Transcripts
NM_001198608.1 NM_001198609.1 NM_001198611.1 NM_001198614.1 NM_001198615.1 NM_001198616.1 NM_001198617.1 NM_001198618.1 NM_001198619.1 NM_003980.4 XM_006715598.2 XM_006715599.2 XM_006715600.2 XM_006715601.3 XM_011536243.2 XM_011536244.2 XM_011536245.1 XM_011536246.2 XM_017011471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
272923 c.1516G>T p.E506* Chr.6: 136362460-136362460 on Build GRCh38
1073687 c.1462C>T p.R488W Chr.6: 136362514-136362514 on Build GRCh38
4839267 c.1407T>G p.T469T Chr.6: 136362569-136362569 on Build GRCh38
6372423 c.1474G>T p.E492* Chr.6: 136362502-136362502 on Build GRCh38
6372422 c.1540G>T p.E514* Chr.6: 136362502-136362502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763919442 Chr.6: 136362473-136362473 on Build GRCh38
rs749870698 Chr.6: 136362525-136362525 on Build GRCh38
rs868502218 Chr.6: 136362495-136362495 on Build GRCh38
rs991369417 Chr.6: 136362381-136362381 on Build GRCh38
rs767523223 Chr.6: 136362456-136362456 on Build GRCh38
rs775741586 Chr.6: 136362548-136362548 on Build GRCh38
rs200026879 Chr.6: 136362428-136362429 on Build GRCh38
rs916690264 Chr.6: 136362382-136362382 on Build GRCh38
rs773576519 Chr.6: 136362516-136362516 on Build GRCh38
rs143028749 Chr.6: 136362513-136362513 on Build GRCh38
rs935290820 Chr.6: 136362424-136362424 on Build GRCh38
rs752419944 Chr.6: 136362425-136362425 on Build GRCh38
rs751772729 Chr.6: 136362431-136362431 on Build GRCh38
rs756261293 Chr.6: 136362470-136362470 on Build GRCh38
rs1046401655 Chr.6: 136362410-136362410 on Build GRCh38
rs771130860 Chr.6: 136362529-136362529 on Build GRCh38
rs767700691 Chr.6: 136362545-136362545 on Build GRCh38
rs200896450 Chr.6: 136362443-136362443 on Build GRCh38
rs752566834 Chr.6: 136362414-136362414 on Build GRCh38
rs553450847 Chr.6: 136362364-136362364 on Build GRCh38
rs763203933 Chr.6: 136362414-136362414 on Build GRCh38
rs952845034 Chr.6: 136362372-136362372 on Build GRCh38
rs778856615 Chr.6: 136362500-136362500 on Build GRCh38
rs766822870 Chr.6: 136362430-136362430 on Build GRCh38
rs777223851 Chr.6: 136362533-136362533 on Build GRCh38
rs758146716 Chr.6: 136362502-136362502 on Build GRCh38
rs1026963358 Chr.6: 136362392-136362392 on Build GRCh38
rs757000885 Chr.6: 136362490-136362490 on Build GRCh38
rs753874599 Chr.6: 136362481-136362481 on Build GRCh38
rs368041365 Chr.6: 136362399-136362399 on Build GRCh38
rs759701304 Chr.6: 136362544-136362544 on Build GRCh38
rs746470141 Chr.6: 136362504-136362504 on Build GRCh38
rs573568745 Chr.6: 136362395-136362395 on Build GRCh38
rs764303837 Chr.6: 136362554-136362554 on Build GRCh38
rs767571413 Chr.6: 136362446-136362446 on Build GRCh38
rs1004327500 Chr.6: 136362401-136362401 on Build GRCh38
rs745460628 Chr.6: 136362501-136362501 on Build GRCh38
rs752809449 Chr.6: 136362467-136362467 on Build GRCh38
rs971002438 Chr.6: 136362493-136362493 on Build GRCh38
rs760661282 Chr.6: 136362553-136362553 on Build GRCh38

View Full Product Details