Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86466324-86466597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373326 c.1174C>G p.Q392E Chr.2: 86466538-86466538 on Build GRCh38
5638689 c.1107C>G p.L369L Chr.2: 86466471-86466471 on Build GRCh38
7373324 c.1120C>G p.Q374E Chr.2: 86466484-86466484 on Build GRCh38
6692350 c.1169G>A p.C390Y Chr.2: 86466533-86466533 on Build GRCh38
4999475 c.1039T>C p.S347P Chr.2: 86466403-86466403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998199542 Chr.2: 86466462-86466462 on Build GRCh38
rs753560307 Chr.2: 86466486-86466486 on Build GRCh38
rs770192890 Chr.2: 86466430-86466430 on Build GRCh38
rs766207623 Chr.2: 86466477-86466477 on Build GRCh38
rs780571362 Chr.2: 86466545-86466545 on Build GRCh38
rs984986054 Chr.2: 86466496-86466496 on Build GRCh38
rs147819585 Chr.2: 86466449-86466449 on Build GRCh38
rs756554104 Chr.2: 86466380-86466380 on Build GRCh38
rs746092509 Chr.2: 86466542-86466542 on Build GRCh38
rs777831801 Chr.2: 86466405-86466405 on Build GRCh38
rs375678230 Chr.2: 86466460-86466460 on Build GRCh38
rs749630885 Chr.2: 86466551-86466551 on Build GRCh38
rs747055924 Chr.2: 86466409-86466409 on Build GRCh38
rs756974200 Chr.2: 86466369-86466369 on Build GRCh38
rs769336391 Chr.2: 86466553-86466553 on Build GRCh38
rs754848773 Chr.2: 86466396-86466396 on Build GRCh38
rs144371910 Chr.2: 86466390-86466390 on Build GRCh38
rs751690769 Chr.2: 86466511-86466511 on Build GRCh38
rs1018820043 Chr.2: 86466392-86466392 on Build GRCh38
rs771597032 Chr.2: 86466566-86466566 on Build GRCh38
rs751056255 Chr.2: 86466362-86466362 on Build GRCh38
rs753268629 Chr.2: 86466356-86466356 on Build GRCh38
rs750083352 Chr.2: 86466469-86466469 on Build GRCh38
rs781337683 Chr.2: 86466527-86466527 on Build GRCh38
rs778777353 Chr.2: 86466506-86466506 on Build GRCh38
rs757331698 Chr.2: 86466515-86466515 on Build GRCh38
rs1007527314 Chr.2: 86466391-86466391 on Build GRCh38
rs750232981 Chr.2: 86466376-86466376 on Build GRCh38
rs761294712 Chr.2: 86466562-86466562 on Build GRCh38
rs746261734 Chr.2: 86466425-86466425 on Build GRCh38
rs760442746 Chr.2: 86466475-86466475 on Build GRCh38
rs200135960 Chr.2: 86466412-86466412 on Build GRCh38
rs765716118 Chr.2: 86466354-86466354 on Build GRCh38
rs755905015 Chr.2: 86466384-86466384 on Build GRCh38
rs370857741 Chr.2: 86466435-86466435 on Build GRCh38
rs372801997 Chr.2: 86466421-86466421 on Build GRCh38
rs147565471 Chr.2: 86466530-86466530 on Build GRCh38
rs753759781 Chr.2: 86466393-86466393 on Build GRCh38
rs769852046 Chr.2: 86466543-86466543 on Build GRCh38
rs761363467 Chr.2: 86466464-86466464 on Build GRCh38
rs767113988 Chr.2: 86466467-86466467 on Build GRCh38
rs778552454 Chr.2: 86466459-86466459 on Build GRCh38
rs200859016 Chr.2: 86466350-86466350 on Build GRCh38
rs772536173 Chr.2: 86466567-86466567 on Build GRCh38
rs369600731 Chr.2: 86466373-86466373 on Build GRCh38
rs774967658 Chr.2: 86466559-86466559 on Build GRCh38
rs546807787 Chr.2: 86466493-86466493 on Build GRCh38
rs763685459 Chr.2: 86466361-86466361 on Build GRCh38
rs762621002 Chr.2: 86466358-86466358 on Build GRCh38
rs774998803 Chr.2: 86466379-86466382 on Build GRCh38
rs993305252 Chr.2: 86466418-86466418 on Build GRCh38
rs760376380 Chr.2: 86466568-86466568 on Build GRCh38
rs190266151 Chr.2: 86466433-86466433 on Build GRCh38

View Full Product Details