Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130762967-130763222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER3
Entrez Gene ID
205147
Gene Name
APC membrane recruitment protein 3
Gene Aliases
FAM123C
UniGene
Hs.369289
Gene Chromosome Location
Chr.2: 130754475-130768134 on Build GRCh38
Transcripts
NM_001105193.1 NM_001105194.1 NM_001105195.1 NM_152698.2 XM_011510793.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3566426 c.1131C>T p.P377P Chr.2: 130763203-130763203 on Build GRCh38
6509751 c.1038G>A p.G346G Chr.2: 130763110-130763110 on Build GRCh38
3566424 c.1083C>T p.G361G Chr.2: 130763155-130763155 on Build GRCh38
6509749 c.895G>A p.A299T Chr.2: 130762967-130762967 on Build GRCh38
3566422 c.1002G>A p.G334G Chr.2: 130763074-130763074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190739558 Chr.2: 130763009-130763009 on Build GRCh38
rs952410383 Chr.2: 130763202-130763202 on Build GRCh38
rs1042232244 Chr.2: 130763097-130763097 on Build GRCh38
rs755473917 Chr.2: 130763027-130763029 on Build GRCh38
rs770899762 Chr.2: 130763172-130763172 on Build GRCh38
rs760092504 Chr.2: 130763174-130763174 on Build GRCh38
rs768384197 Chr.2: 130763091-130763091 on Build GRCh38
rs755323037 Chr.2: 130763054-130763054 on Build GRCh38
rs773236761 Chr.2: 130763169-130763169 on Build GRCh38
rs866346930 Chr.2: 130763185-130763185 on Build GRCh38
rs747211046 Chr.2: 130763017-130763017 on Build GRCh38
rs555917912 Chr.2: 130763114-130763114 on Build GRCh38
rs775061865 Chr.2: 130763173-130763173 on Build GRCh38
rs551366921 Chr.2: 130763015-130763015 on Build GRCh38
rs927688504 Chr.2: 130763108-130763108 on Build GRCh38
rs749984864 Chr.2: 130763133-130763133 on Build GRCh38
rs868833929 Chr.2: 130763044-130763044 on Build GRCh38
rs186201647 Chr.2: 130762993-130762993 on Build GRCh38
rs770084821 Chr.2: 130763167-130763167 on Build GRCh38
rs778457608 Chr.2: 130763071-130763071 on Build GRCh38
rs983772806 Chr.2: 130763052-130763052 on Build GRCh38
rs555251292 Chr.2: 130763093-130763093 on Build GRCh38
rs113802956 Chr.2: 130763024-130763024 on Build GRCh38
rs756898204 Chr.2: 130763068-130763068 on Build GRCh38
rs753138909 Chr.2: 130763181-130763181 on Build GRCh38
rs772073367 Chr.2: 130763086-130763086 on Build GRCh38
rs994290983 Chr.2: 130763022-130763022 on Build GRCh38
rs976919134 Chr.2: 130763088-130763088 on Build GRCh38
rs763102634 Chr.2: 130763171-130763171 on Build GRCh38
rs1028275575 Chr.2: 130763034-130763034 on Build GRCh38
rs753588139 Chr.2: 130763126-130763126 on Build GRCh38
rs750932360 Chr.2: 130763030-130763030 on Build GRCh38
rs200166860 Chr.2: 130763105-130763105 on Build GRCh38
rs745845674 Chr.2: 130763073-130763073 on Build GRCh38
rs766746775 Chr.2: 130763045-130763045 on Build GRCh38
rs775251651 Chr.2: 130763087-130763087 on Build GRCh38
rs751981550 Chr.2: 130763053-130763053 on Build GRCh38
rs774210011 Chr.2: 130762996-130762996 on Build GRCh38
rs371693870 Chr.2: 130763110-130763110 on Build GRCh38
rs530043742 Chr.2: 130763158-130763158 on Build GRCh38
rs757781233 Chr.2: 130763137-130763137 on Build GRCh38
rs537694206 Chr.2: 130763113-130763113 on Build GRCh38
rs148342332 Chr.2: 130763203-130763203 on Build GRCh38
rs1905235 Chr.2: 130763090-130763090 on Build GRCh38
rs745874912 Chr.2: 130763099-130763099 on Build GRCh38
rs767986216 Chr.2: 130763117-130763117 on Build GRCh38
rs371197985 Chr.2: 130763196-130763196 on Build GRCh38
rs780940947 Chr.2: 130763157-130763157 on Build GRCh38
rs761990427 Chr.2: 130763094-130763094 on Build GRCh38
rs777579863 Chr.2: 130763063-130763063 on Build GRCh38
rs371270046 Chr.2: 130763016-130763016 on Build GRCh38
rs368379439 Chr.2: 130763037-130763037 on Build GRCh38
rs778862261 Chr.2: 130763152-130763152 on Build GRCh38
rs202240427 Chr.2: 130763194-130763194 on Build GRCh38
rs761084545 Chr.2: 130763191-130763191 on Build GRCh38
rs780054373 Chr.2: 130763148-130763148 on Build GRCh38
rs749986141 Chr.2: 130763195-130763195 on Build GRCh38
rs200015684 Chr.2: 130763025-130763025 on Build GRCh38
rs1055694945 Chr.2: 130763106-130763106 on Build GRCh38
rs372383192 Chr.2: 130763067-130763067 on Build GRCh38
rs746427562 Chr.2: 130763129-130763129 on Build GRCh38
rs768045393 Chr.2: 130763178-130763178 on Build GRCh38
rs367830628 Chr.2: 130763153-130763153 on Build GRCh38
rs766910809 Chr.2: 130763111-130763111 on Build GRCh38
rs776767192 Chr.2: 130763021-130763021 on Build GRCh38
rs539498441 Chr.2: 130763008-130763008 on Build GRCh38

View Full Product Details