Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142775290-142775515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYNX1
Entrez Gene ID
66004
Gene Name
Ly6/neurotoxin 1
Gene Aliases
SLURP2
UniGene
Hs.158665
Gene Chromosome Location
Chr.8: 142764334-142777848 on Build GRCh38
Transcripts
NM_023946.3 NM_177457.4 NM_177476.3 NM_177477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698688 c.218A>G p.Y73C Chr.8: 142775300-142775300 on Build GRCh38
1151045 c.200G>T p.R67L Chr.8: 142775318-142775318 on Build GRCh38
3897329 c.192C>T p.C64C Chr.8: 142775326-142775326 on Build GRCh38
6536657 c.210G>C p.E70D Chr.8: 142775308-142775308 on Build GRCh38
3897328 c.225C>T p.G75G Chr.8: 142775293-142775293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587747914 Chr.8: 142775450-142775450 on Build GRCh38
rs753961289 Chr.8: 142775357-142775357 on Build GRCh38
rs769784395 Chr.8: 142775381-142775381 on Build GRCh38
rs1054481062 Chr.8: 142775429-142775429 on Build GRCh38
rs587631495 Chr.8: 142775411-142775411 on Build GRCh38
rs587676053 Chr.8: 142775463-142775463 on Build GRCh38
rs773681151 Chr.8: 142775402-142775402 on Build GRCh38
rs192474652 Chr.8: 142775413-142775413 on Build GRCh38
rs774106728 Chr.8: 142775328-142775328 on Build GRCh38
rs770820982 Chr.8: 142775325-142775325 on Build GRCh38
rs373239191 Chr.8: 142775322-142775322 on Build GRCh38
rs893973855 Chr.8: 142775432-142775432 on Build GRCh38
rs867535838 Chr.8: 142775330-142775330 on Build GRCh38
rs374446309 Chr.8: 142775394-142775394 on Build GRCh38
rs114034813 Chr.8: 142775342-142775342 on Build GRCh38
rs367723215 Chr.8: 142775395-142775395 on Build GRCh38
rs587598163 Chr.8: 142775456-142775456 on Build GRCh38
rs758591221 Chr.8: 142775378-142775378 on Build GRCh38
rs766779595 Chr.8: 142775355-142775355 on Build GRCh38
rs766909026 Chr.8: 142775407-142775407 on Build GRCh38
rs993463510 Chr.8: 142775399-142775399 on Build GRCh38
rs779133253 Chr.8: 142775358-142775358 on Build GRCh38
rs1010154481 Chr.8: 142775451-142775451 on Build GRCh38
rs765344128 Chr.8: 142775412-142775412 on Build GRCh38
rs761308415 Chr.8: 142775347-142775347 on Build GRCh38
rs201126134 Chr.8: 142775321-142775321 on Build GRCh38
rs376146344 Chr.8: 142775375-142775375 on Build GRCh38
rs199529658 Chr.8: 142775318-142775318 on Build GRCh38
rs182621790 Chr.8: 142775319-142775319 on Build GRCh38
rs752968943 Chr.8: 142775371-142775371 on Build GRCh38
rs746927308 Chr.8: 142775376-142775376 on Build GRCh38
rs587733724 Chr.8: 142775409-142775409 on Build GRCh38
rs377016634 Chr.8: 142775400-142775400 on Build GRCh38
rs371093337 Chr.8: 142775380-142775380 on Build GRCh38
rs980541743 Chr.8: 142775474-142775474 on Build GRCh38
rs763843306 Chr.8: 142775326-142775326 on Build GRCh38
rs750720051 Chr.8: 142775344-142775344 on Build GRCh38
rs972119885 Chr.8: 142775350-142775350 on Build GRCh38
rs761595613 Chr.8: 142775396-142775396 on Build GRCh38

View Full Product Details