Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46350172-46350399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0226L
Entrez Gene ID
80183
Gene Name
KIAA0226 like
Gene Aliases
C13orf18
UniGene
Hs.98117
Gene Chromosome Location
Chr.13: 46342000-46390042 on Build GRCh38
Transcripts
NM_001286761.1 NM_001286762.1 NM_001286763.1 NM_001286764.1 NM_001286765.1 NM_001286766.1 NM_025113.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864680 c.1205C>T p.S402L Chr.13: 46350276-46350276 on Build GRCh38
1246441 c.1456G>A p.V486I Chr.13: 46350226-46350226 on Build GRCh38
218438 c.1343G>A p.R448Q Chr.13: 46350339-46350339 on Build GRCh38
4215233 c.1407G>A p.S469S Chr.13: 46350275-46350275 on Build GRCh38
3716417 c.1420C>T p.R474* Chr.13: 46350262-46350262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373829399 Chr.13: 46350265-46350265 on Build GRCh38
rs763412476 Chr.13: 46350208-46350208 on Build GRCh38
rs755996619 Chr.13: 46350261-46350261 on Build GRCh38
rs200810574 Chr.13: 46350325-46350325 on Build GRCh38
rs1040797485 Chr.13: 46350229-46350229 on Build GRCh38
rs368540256 Chr.13: 46350199-46350199 on Build GRCh38
rs772688546 Chr.13: 46350340-46350340 on Build GRCh38
rs749602713 Chr.13: 46350331-46350331 on Build GRCh38
rs371701895 Chr.13: 46350276-46350276 on Build GRCh38
rs760220192 Chr.13: 46350359-46350359 on Build GRCh38
rs776130397 Chr.13: 46350295-46350295 on Build GRCh38
rs371814111 Chr.13: 46350200-46350200 on Build GRCh38
rs1006439734 Chr.13: 46350291-46350291 on Build GRCh38
rs745989558 Chr.13: 46350285-46350285 on Build GRCh38
rs763648996 Chr.13: 46350210-46350210 on Build GRCh38
rs901151102 Chr.13: 46350355-46350355 on Build GRCh38
rs781370422 Chr.13: 46350279-46350279 on Build GRCh38
rs866599289 Chr.13: 46350327-46350327 on Build GRCh38
rs138546921 Chr.13: 46350228-46350228 on Build GRCh38
rs766389615 Chr.13: 46350262-46350262 on Build GRCh38
rs368013479 Chr.13: 46350375-46350375 on Build GRCh38
rs747173963 Chr.13: 46350275-46350275 on Build GRCh38
rs149265486 Chr.13: 46350339-46350339 on Build GRCh38
rs764133902 Chr.13: 46350316-46350316 on Build GRCh38
rs774582818 Chr.13: 46350334-46350334 on Build GRCh38
rs147137455 Chr.13: 46350227-46350227 on Build GRCh38
rs139297008 Chr.13: 46350198-46350198 on Build GRCh38
rs778518328 Chr.13: 46350365-46350365 on Build GRCh38
rs888506175 Chr.13: 46350337-46350337 on Build GRCh38
rs201645593 Chr.13: 46350272-46350272 on Build GRCh38
rs143465179 Chr.13: 46350219-46350219 on Build GRCh38
rs754872901 Chr.13: 46350271-46350271 on Build GRCh38
rs775691381 Chr.13: 46350205-46350205 on Build GRCh38
rs565696372 Chr.13: 46350366-46350366 on Build GRCh38
rs753678651 Chr.13: 46350333-46350333 on Build GRCh38
rs367676102 Chr.13: 46350373-46350373 on Build GRCh38
rs552737686 Chr.13: 46350220-46350220 on Build GRCh38
rs770389783 Chr.13: 46350290-46350290 on Build GRCh38
rs749969162 Chr.13: 46350237-46350237 on Build GRCh38

View Full Product Details