Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43396719-43396984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_011515225.2 XM_011515226.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3230199 c.584_587delCCGT p.V196fs*8 Chr.7: 43396842-43396845 on Build GRCh38
746877 c.600A>G p.G200G Chr.7: 43396858-43396858 on Build GRCh38
4689916 c.598G>T p.G200* Chr.7: 43396856-43396856 on Build GRCh38
1313088 c.631+1G>C p.? Chr.7: 43396890-43396890 on Build GRCh38
1755274 c.630A>T p.S210S Chr.7: 43396888-43396888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547939909 Chr.7: 43396791-43396791 on Build GRCh38
rs775172566 Chr.7: 43396930-43396930 on Build GRCh38
rs745473483 Chr.7: 43396946-43396947 on Build GRCh38
rs776309267 Chr.7: 43396777-43396777 on Build GRCh38
rs764872831 Chr.7: 43396824-43396824 on Build GRCh38
rs773344957 Chr.7: 43396766-43396766 on Build GRCh38
rs762506249 Chr.7: 43396827-43396827 on Build GRCh38
rs763697168 Chr.7: 43396932-43396932 on Build GRCh38
rs750980231 Chr.7: 43396842-43396842 on Build GRCh38
rs776563545 Chr.7: 43396905-43396905 on Build GRCh38
rs915229925 Chr.7: 43396952-43396952 on Build GRCh38
rs770859134 Chr.7: 43396776-43396776 on Build GRCh38
rs772331590 Chr.7: 43396867-43396867 on Build GRCh38
rs770785112 Chr.7: 43396903-43396903 on Build GRCh38
rs375753925 Chr.7: 43396870-43396870 on Build GRCh38
rs373256741 Chr.7: 43396920-43396920 on Build GRCh38
rs190590512 Chr.7: 43396902-43396902 on Build GRCh38
rs759315657 Chr.7: 43396784-43396784 on Build GRCh38
rs1026608534 Chr.7: 43396789-43396789 on Build GRCh38
rs752341096 Chr.7: 43396825-43396825 on Build GRCh38
rs755329283 Chr.7: 43396847-43396847 on Build GRCh38
rs763572809 Chr.7: 43396830-43396830 on Build GRCh38
rs762561514 Chr.7: 43396931-43396931 on Build GRCh38
rs374959981 Chr.7: 43396843-43396843 on Build GRCh38
rs201771274 Chr.7: 43396844-43396844 on Build GRCh38
rs750030710 Chr.7: 43396923-43396924 on Build GRCh38
rs989995451 Chr.7: 43396875-43396875 on Build GRCh38
rs779347290 Chr.7: 43396863-43396863 on Build GRCh38
rs760493961 Chr.7: 43396772-43396772 on Build GRCh38
rs748371744 Chr.7: 43396866-43396866 on Build GRCh38
rs759314493 Chr.7: 43396909-43396909 on Build GRCh38

View Full Product Details