Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116792304-116792563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRC6A
Entrez Gene ID
222545
Gene Name
G protein-coupled receptor class C group 6 member A
Gene Aliases
GPCR, bA86F4.3
UniGene
Hs.266745
Gene Chromosome Location
Chr.6: 116788512-116831128 on Build GRCh38
Transcripts
NM_001286354.1 NM_001286355.1 NM_148963.3 XM_017010475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173346 c.2501G>T p.C834F Chr.6: 116792422-116792422 on Build GRCh38
4538871 c.2613G>A p.L871L Chr.6: 116792310-116792310 on Build GRCh38
6489285 c.2602C>G p.P868A Chr.6: 116792321-116792321 on Build GRCh38
6547595 c.2475C>A p.I825I Chr.6: 116792448-116792448 on Build GRCh38
1072457 c.2370T>G p.I790M Chr.6: 116792553-116792553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747253095 Chr.6: 116792470-116792470 on Build GRCh38
rs763381094 Chr.6: 116792502-116792502 on Build GRCh38
rs767205030 Chr.6: 116792526-116792526 on Build GRCh38
rs770426370 Chr.6: 116792413-116792413 on Build GRCh38
rs945249771 Chr.6: 116792481-116792481 on Build GRCh38
rs615199 Chr.6: 116792490-116792490 on Build GRCh38
rs750092727 Chr.6: 116792332-116792332 on Build GRCh38
rs753583086 Chr.6: 116792456-116792456 on Build GRCh38
rs762476414 Chr.6: 116792406-116792406 on Build GRCh38
rs752631270 Chr.6: 116792446-116792446 on Build GRCh38
rs906562085 Chr.6: 116792420-116792420 on Build GRCh38
rs777182062 Chr.6: 116792403-116792403 on Build GRCh38
rs751684558 Chr.6: 116792457-116792457 on Build GRCh38
rs774043932 Chr.6: 116792513-116792513 on Build GRCh38
rs773649536 Chr.6: 116792418-116792418 on Build GRCh38
rs373924677 Chr.6: 116792359-116792359 on Build GRCh38
rs762905959 Chr.6: 116792385-116792385 on Build GRCh38
rs749087006 Chr.6: 116792390-116792390 on Build GRCh38
rs765958789 Chr.6: 116792344-116792344 on Build GRCh38
rs367740558 Chr.6: 116792422-116792422 on Build GRCh38
rs201038107 Chr.6: 116792370-116792370 on Build GRCh38
rs759181148 Chr.6: 116792520-116792520 on Build GRCh38
rs759067820 Chr.6: 116792426-116792426 on Build GRCh38
rs751360329 Chr.6: 116792455-116792458 on Build GRCh38
rs751028477 Chr.6: 116792369-116792369 on Build GRCh38
rs1043659669 Chr.6: 116792484-116792484 on Build GRCh38
rs773884316 Chr.6: 116792496-116792496 on Build GRCh38
rs777613092 Chr.6: 116792381-116792381 on Build GRCh38
rs758052885 Chr.6: 116792339-116792339 on Build GRCh38
rs757098502 Chr.6: 116792466-116792468 on Build GRCh38
rs770373383 Chr.6: 116792493-116792493 on Build GRCh38
rs755894078 Chr.6: 116792379-116792379 on Build GRCh38
rs1053931715 Chr.6: 116792346-116792346 on Build GRCh38
rs747079091 Chr.6: 116792398-116792398 on Build GRCh38
rs778258625 Chr.6: 116792445-116792445 on Build GRCh38
rs566847270 Chr.6: 116792334-116792334 on Build GRCh38
rs751214810 Chr.6: 116792425-116792425 on Build GRCh38
rs781155252 Chr.6: 116792371-116792371 on Build GRCh38
rs1028040985 Chr.6: 116792353-116792353 on Build GRCh38
rs766999051 Chr.6: 116792441-116792441 on Build GRCh38
rs767181113 Chr.6: 116792508-116792512 on Build GRCh38
rs750666167 Chr.6: 116792529-116792529 on Build GRCh38
rs781062631 Chr.6: 116792488-116792488 on Build GRCh38
rs1049938747 Chr.6: 116792527-116792527 on Build GRCh38
rs367778621 Chr.6: 116792402-116792402 on Build GRCh38
rs748089208 Chr.6: 116792373-116792373 on Build GRCh38
rs763430536 Chr.6: 116792421-116792421 on Build GRCh38
rs142053954 Chr.6: 116792395-116792395 on Build GRCh38
rs768775607 Chr.6: 116792485-116792485 on Build GRCh38
rs755980101 Chr.6: 116792447-116792447 on Build GRCh38
rs764063363 Chr.6: 116792368-116792369 on Build GRCh38
rs777781673 Chr.6: 116792491-116792491 on Build GRCh38
rs151151248 Chr.6: 116792451-116792451 on Build GRCh38

View Full Product Details