Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3092383-3092634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSBPL5
Entrez Gene ID
114879
Gene Name
oxysterol binding protein like 5
Gene Aliases
OBPH1, ORP5
UniGene
Hs.436166
Gene Chromosome Location
Chr.11: 3087116-3165352 on Build GRCh38
Transcripts
NM_001144063.1 NM_020896.3 NM_145638.2 XM_011519873.2 XM_017017163.1 XM_017017164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449406 c.2212_2214delGTG p.V738delV Chr.11: 3092477-3092479 on Build GRCh38
4446759 c.2133-3C>T p.? Chr.11: 3092561-3092561 on Build GRCh38
5512737 c.2252G>T p.R751M Chr.11: 3092439-3092439 on Build GRCh38
4535646 c.2212G>T p.V738L Chr.11: 3092479-3092479 on Build GRCh38
6421272 c.2155A>G p.K719E Chr.11: 3092536-3092536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890515048 Chr.11: 3092456-3092456 on Build GRCh38
rs934323617 Chr.11: 3092589-3092589 on Build GRCh38
rs376315376 Chr.11: 3092435-3092435 on Build GRCh38
rs371043968 Chr.11: 3092420-3092420 on Build GRCh38
rs963446704 Chr.11: 3092513-3092513 on Build GRCh38
rs764398770 Chr.11: 3092531-3092531 on Build GRCh38
rs771435217 Chr.11: 3092461-3092461 on Build GRCh38
rs979863733 Chr.11: 3092597-3092597 on Build GRCh38
rs151250529 Chr.11: 3092473-3092473 on Build GRCh38
rs371190248 Chr.11: 3092439-3092439 on Build GRCh38
rs753781092 Chr.11: 3092576-3092576 on Build GRCh38
rs751956572 Chr.11: 3092545-3092545 on Build GRCh38
rs1009655528 Chr.11: 3092452-3092452 on Build GRCh38
rs906881128 Chr.11: 3092418-3092418 on Build GRCh38
rs757977452 Chr.11: 3092607-3092607 on Build GRCh38
rs771801579 Chr.11: 3092510-3092510 on Build GRCh38
rs778029339 Chr.11: 3092404-3092404 on Build GRCh38
rs749151635 Chr.11: 3092437-3092437 on Build GRCh38
rs754997813 Chr.11: 3092583-3092583 on Build GRCh38
rs781101351 Chr.11: 3092482-3092482 on Build GRCh38
rs770200561 Chr.11: 3092525-3092525 on Build GRCh38
rs138015375 Chr.11: 3092509-3092509 on Build GRCh38
rs374070869 Chr.11: 3092472-3092472 on Build GRCh38
rs1045445785 Chr.11: 3092411-3092411 on Build GRCh38
rs759933628 Chr.11: 3092465-3092465 on Build GRCh38
rs763470706 Chr.11: 3092528-3092528 on Build GRCh38
rs781350724 Chr.11: 3092429-3092429 on Build GRCh38
rs150434379 Chr.11: 3092498-3092498 on Build GRCh38
rs764453800 Chr.11: 3092475-3092475 on Build GRCh38
rs747233805 Chr.11: 3092419-3092419 on Build GRCh38
rs768514846 Chr.11: 3092499-3092499 on Build GRCh38
rs748028696 Chr.11: 3092603-3092603 on Build GRCh38
rs1011740489 Chr.11: 3092455-3092455 on Build GRCh38
rs1052784379 Chr.11: 3092601-3092601 on Build GRCh38
rs760292668 Chr.11: 3092511-3092511 on Build GRCh38
rs750564557 Chr.11: 3092484-3092484 on Build GRCh38
rs774082217 Chr.11: 3092445-3092445 on Build GRCh38
rs112904220 Chr.11: 3092609-3092609 on Build GRCh38
rs143610110 Chr.11: 3092547-3092547 on Build GRCh38
rs751727731 Chr.11: 3092479-3092479 on Build GRCh38
rs111371897 Chr.11: 3092489-3092489 on Build GRCh38
rs371607503 Chr.11: 3092587-3092587 on Build GRCh38
rs141473758 Chr.11: 3092453-3092453 on Build GRCh38
rs778874744 Chr.11: 3092500-3092500 on Build GRCh38
rs371170315 Chr.11: 3092480-3092480 on Build GRCh38
rs892721622 Chr.11: 3092450-3092450 on Build GRCh38
rs756154711 Chr.11: 3092563-3092563 on Build GRCh38
rs374457287 Chr.11: 3092434-3092434 on Build GRCh38
rs772814832 Chr.11: 3092463-3092463 on Build GRCh38
rs367958562 Chr.11: 3092562-3092562 on Build GRCh38
rs970319525 Chr.11: 3092523-3092523 on Build GRCh38
rs928535126 Chr.11: 3092577-3092577 on Build GRCh38
rs780205096 Chr.11: 3092491-3092491 on Build GRCh38
rs780295645 Chr.11: 3092568-3092568 on Build GRCh38
rs767837222 Chr.11: 3092557-3092557 on Build GRCh38
rs141858255 Chr.11: 3092466-3092466 on Build GRCh38

View Full Product Details