Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34860113-34860366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_017027809.1 XM_017027810.1 XM_017027811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5343580 c.727C>T p.H243Y Chr.20: 34860270-34860270 on Build GRCh38
3693448 c.737A>G p.Y246C Chr.20: 34860260-34860260 on Build GRCh38
4820302 c.701G>A p.G234E Chr.20: 34860296-34860296 on Build GRCh38
4794924 c.676-1G>A p.? Chr.20: 34860322-34860322 on Build GRCh38
6273367 c.677C>A p.P226H Chr.20: 34860320-34860320 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6060095 Chr.20: 34860286-34860286 on Build GRCh38
rs764688866 Chr.20: 34860298-34860298 on Build GRCh38
rs774771343 Chr.20: 34860310-34860310 on Build GRCh38
rs753993169 Chr.20: 34860340-34860340 on Build GRCh38
rs756418112 Chr.20: 34860333-34860333 on Build GRCh38
rs968513634 Chr.20: 34860171-34860171 on Build GRCh38
rs774875253 Chr.20: 34860222-34860223 on Build GRCh38
rs979542803 Chr.20: 34860178-34860178 on Build GRCh38
rs919789984 Chr.20: 34860301-34860301 on Build GRCh38
rs781600276 Chr.20: 34860215-34860215 on Build GRCh38
rs768745642 Chr.20: 34860224-34860224 on Build GRCh38
rs372362464 Chr.20: 34860260-34860260 on Build GRCh38
rs750652638 Chr.20: 34860216-34860216 on Build GRCh38
rs764521693 Chr.20: 34860208-34860208 on Build GRCh38
rs778665554 Chr.20: 34860229-34860229 on Build GRCh38
rs1002395332 Chr.20: 34860192-34860194 on Build GRCh38
rs6142253 Chr.20: 34860338-34860338 on Build GRCh38
rs140622797 Chr.20: 34860262-34860262 on Build GRCh38
rs181027490 Chr.20: 34860329-34860329 on Build GRCh38
rs750788716 Chr.20: 34860332-34860332 on Build GRCh38
rs762354131 Chr.20: 34860325-34860325 on Build GRCh38
rs573361809 Chr.20: 34860182-34860182 on Build GRCh38
rs956737987 Chr.20: 34860198-34860198 on Build GRCh38
rs766643384 Chr.20: 34860334-34860334 on Build GRCh38
rs558000732 Chr.20: 34860175-34860175 on Build GRCh38
rs780340344 Chr.20: 34860220-34860220 on Build GRCh38
rs776119649 Chr.20: 34860293-34860293 on Build GRCh38
rs759010368 Chr.20: 34860297-34860297 on Build GRCh38
rs757517380 Chr.20: 34860214-34860214 on Build GRCh38
rs746719350 Chr.20: 34860271-34860271 on Build GRCh38
rs770508867 Chr.20: 34860283-34860283 on Build GRCh38
rs376736314 Chr.20: 34860200-34860200 on Build GRCh38
rs748094426 Chr.20: 34860252-34860252 on Build GRCh38
rs749322972 Chr.20: 34860222-34860222 on Build GRCh38
rs879023714 Chr.20: 34860314-34860314 on Build GRCh38
rs763460437 Chr.20: 34860207-34860207 on Build GRCh38
rs201952715 Chr.20: 34860345-34860345 on Build GRCh38
rs115343976 Chr.20: 34860213-34860213 on Build GRCh38
rs369384619 Chr.20: 34860219-34860219 on Build GRCh38
rs8183445 Chr.20: 34860238-34860238 on Build GRCh38

View Full Product Details