Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 155436381-155436622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOX3
Entrez Gene ID
50508
Gene Name
NADPH oxidase 3
Gene Aliases
GP91-3, MOX-2
UniGene
Hs.247776
Gene Chromosome Location
Chr.6: 155395368-155455903 on Build GRCh38
Transcripts
NM_015718.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5723864 c.769G>T p.V257L Chr.6: 155436447-155436447 on Build GRCh38
4508172 c.767C>T p.P256L Chr.6: 155436449-155436449 on Build GRCh38
3022172 c.725G>T p.C242F Chr.6: 155436491-155436491 on Build GRCh38
3622248 c.768C>T p.P256P Chr.6: 155436448-155436448 on Build GRCh38
1075333 c.752C>A p.T251K Chr.6: 155436464-155436464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913878379 Chr.6: 155436520-155436520 on Build GRCh38
rs781326794 Chr.6: 155436583-155436584 on Build GRCh38
rs79594480 Chr.6: 155436531-155436531 on Build GRCh38
rs756137259 Chr.6: 155436532-155436532 on Build GRCh38
rs533793580 Chr.6: 155436500-155436500 on Build GRCh38
rs753391396 Chr.6: 155436420-155436420 on Build GRCh38
rs767886132 Chr.6: 155436539-155436539 on Build GRCh38
rs751815031 Chr.6: 155436446-155436446 on Build GRCh38
rs527483156 Chr.6: 155436482-155436482 on Build GRCh38
rs756197732 Chr.6: 155436544-155436544 on Build GRCh38
rs769049081 Chr.6: 155436418-155436418 on Build GRCh38
rs568380489 Chr.6: 155436431-155436431 on Build GRCh38
rs140761547 Chr.6: 155436447-155436447 on Build GRCh38
rs763038269 Chr.6: 155436508-155436508 on Build GRCh38
rs760251192 Chr.6: 155436483-155436483 on Build GRCh38
rs759113179 Chr.6: 155436491-155436491 on Build GRCh38
rs150291307 Chr.6: 155436503-155436503 on Build GRCh38
rs988527008 Chr.6: 155436514-155436514 on Build GRCh38
rs201236143 Chr.6: 155436595-155436595 on Build GRCh38
rs780580625 Chr.6: 155436466-155436466 on Build GRCh38
rs374977580 Chr.6: 155436582-155436582 on Build GRCh38
rs777757925 Chr.6: 155436444-155436444 on Build GRCh38
rs763918734 Chr.6: 155436525-155436525 on Build GRCh38
rs750652267 Chr.6: 155436540-155436540 on Build GRCh38
rs181984491 Chr.6: 155436546-155436546 on Build GRCh38
rs755479061 Chr.6: 155436571-155436571 on Build GRCh38
rs58861026 Chr.6: 155436557-155436557 on Build GRCh38
rs758178198 Chr.6: 155436443-155436443 on Build GRCh38
rs201840949 Chr.6: 155436476-155436476 on Build GRCh38
rs202017556 Chr.6: 155436578-155436578 on Build GRCh38
rs143023863 Chr.6: 155436590-155436590 on Build GRCh38
rs879038171 Chr.6: 155436449-155436449 on Build GRCh38
rs539125619 Chr.6: 155436509-155436509 on Build GRCh38
rs770164038 Chr.6: 155436457-155436457 on Build GRCh38
rs759334602 Chr.6: 155436421-155436421 on Build GRCh38
rs775954849 Chr.6: 155436488-155436488 on Build GRCh38
rs755180954 Chr.6: 155436419-155436419 on Build GRCh38
rs368365169 Chr.6: 155436478-155436478 on Build GRCh38
rs770582653 Chr.6: 155436486-155436486 on Build GRCh38
rs137953478 Chr.6: 155436528-155436528 on Build GRCh38
rs927844894 Chr.6: 155436518-155436518 on Build GRCh38
rs371963780 Chr.6: 155436454-155436454 on Build GRCh38
rs574379568 Chr.6: 155436591-155436591 on Build GRCh38
rs763068128 Chr.6: 155436552-155436552 on Build GRCh38
rs762928252 Chr.6: 155436594-155436594 on Build GRCh38
rs370262095 Chr.6: 155436572-155436572 on Build GRCh38
rs781366402 Chr.6: 155436450-155436450 on Build GRCh38
rs192410865 Chr.6: 155436537-155436537 on Build GRCh38

View Full Product Details