Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141422604-141422843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032091.1 NM_032092.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3409944 c.1478C>T p.T493M Chr.5: 141422705-141422705 on Build GRCh38
3429108 c.1390G>A p.E464K Chr.5: 141422617-141422617 on Build GRCh38
5692136 c.1534A>T p.T512S Chr.5: 141422761-141422761 on Build GRCh38
3852029 c.1558T>A p.S520T Chr.5: 141422785-141422785 on Build GRCh38
351162 c.1429G>C p.A477P Chr.5: 141422656-141422656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758922274 Chr.5: 141422706-141422706 on Build GRCh38
rs868426196 Chr.5: 141422806-141422806 on Build GRCh38
rs373180311 Chr.5: 141422727-141422727 on Build GRCh38
rs778578440 Chr.5: 141422750-141422750 on Build GRCh38
rs748692494 Chr.5: 141422646-141422646 on Build GRCh38
rs199543811 Chr.5: 141422632-141422632 on Build GRCh38
rs770618826 Chr.5: 141422644-141422644 on Build GRCh38
rs369004166 Chr.5: 141422819-141422819 on Build GRCh38
rs746943605 Chr.5: 141422781-141422781 on Build GRCh38
rs759548203 Chr.5: 141422656-141422656 on Build GRCh38
rs560544401 Chr.5: 141422724-141422724 on Build GRCh38
rs540507159 Chr.5: 141422671-141422671 on Build GRCh38
rs754910458 Chr.5: 141422670-141422671 on Build GRCh38
rs557969590 Chr.5: 141422641-141422641 on Build GRCh38
rs1048315821 Chr.5: 141422720-141422721 on Build GRCh38
rs904706749 Chr.5: 141422820-141422820 on Build GRCh38
rs765368737 Chr.5: 141422699-141422699 on Build GRCh38
rs762009100 Chr.5: 141422692-141422692 on Build GRCh38
rs754112462 Chr.5: 141422685-141422685 on Build GRCh38
rs776409955 Chr.5: 141422789-141422789 on Build GRCh38
rs766395950 Chr.5: 141422820-141422821 on Build GRCh38
rs762981102 Chr.5: 141422821-141422821 on Build GRCh38
rs771844166 Chr.5: 141422662-141422662 on Build GRCh38
rs372343628 Chr.5: 141422774-141422774 on Build GRCh38
rs769515606 Chr.5: 141422633-141422633 on Build GRCh38
rs898276821 Chr.5: 141422798-141422798 on Build GRCh38
rs777330051 Chr.5: 141422639-141422639 on Build GRCh38
rs369208205 Chr.5: 141422677-141422677 on Build GRCh38
rs1041081332 Chr.5: 141422778-141422778 on Build GRCh38
rs371027437 Chr.5: 141422705-141422705 on Build GRCh38
rs745320796 Chr.5: 141422762-141422762 on Build GRCh38
rs543001807 Chr.5: 141422795-141422795 on Build GRCh38
rs900827963 Chr.5: 141422792-141422792 on Build GRCh38
rs752071139 Chr.5: 141422722-141422722 on Build GRCh38
rs199507728 Chr.5: 141422807-141422807 on Build GRCh38
rs780383828 Chr.5: 141422708-141422708 on Build GRCh38
rs188587553 Chr.5: 141422655-141422655 on Build GRCh38
rs773990745 Chr.5: 141422647-141422647 on Build GRCh38
rs748292321 Chr.5: 141422758-141422759 on Build GRCh38
rs780472571 Chr.5: 141422629-141422629 on Build GRCh38
rs775331499 Chr.5: 141422779-141422779 on Build GRCh38
rs779043968 Chr.5: 141422678-141422679 on Build GRCh38
rs202046219 Chr.5: 141422749-141422749 on Build GRCh38
rs980166515 Chr.5: 141422637-141422637 on Build GRCh38
rs536737009 Chr.5: 141422740-141422740 on Build GRCh38
rs768397979 Chr.5: 141422783-141422783 on Build GRCh38
rs763386007 Chr.5: 141422746-141422746 on Build GRCh38
rs1037091316 Chr.5: 141422797-141422797 on Build GRCh38
rs931128847 Chr.5: 141422713-141422713 on Build GRCh38
rs748248571 Chr.5: 141422811-141422811 on Build GRCh38
rs775380177 Chr.5: 141422666-141422666 on Build GRCh38

View Full Product Details