Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7847693-7847924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5L
Entrez Gene ID
115704
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
-
UniGene
Hs.26870
Gene Chromosome Location
Chr.19: 7830275-7864976 on Build GRCh38
Transcripts
NM_001159944.2 NM_145245.4 XM_005272458.4 XM_017026239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1003386 c.279C>T p.I93I Chr.19: 7847873-7847873 on Build GRCh38
5365600 c.280G>A p.A94T Chr.19: 7847874-7847874 on Build GRCh38
6365212 c.302G>A p.R101Q Chr.19: 7847896-7847896 on Build GRCh38
1003385 c.138-1G>T p.? Chr.19: 7847731-7847731 on Build GRCh38
79809 c.180C>T p.G60G Chr.19: 7847774-7847774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754679538 Chr.19: 7847850-7847850 on Build GRCh38
rs779624210 Chr.19: 7847898-7847898 on Build GRCh38
rs140398803 Chr.19: 7847852-7847852 on Build GRCh38
rs370414994 Chr.19: 7847778-7847778 on Build GRCh38
rs199527137 Chr.19: 7847741-7847741 on Build GRCh38
rs1007662081 Chr.19: 7847819-7847819 on Build GRCh38
rs764314362 Chr.19: 7847891-7847891 on Build GRCh38
rs749715262 Chr.19: 7847760-7847760 on Build GRCh38
rs756371946 Chr.19: 7847758-7847758 on Build GRCh38
rs761843617 Chr.19: 7847716-7847716 on Build GRCh38
rs868044398 Chr.19: 7847802-7847802 on Build GRCh38
rs753092741 Chr.19: 7847745-7847745 on Build GRCh38
rs771840893 Chr.19: 7847777-7847777 on Build GRCh38
rs1030298860 Chr.19: 7847782-7847782 on Build GRCh38
rs771585408 Chr.19: 7847761-7847761 on Build GRCh38
rs754276273 Chr.19: 7847895-7847895 on Build GRCh38
rs373550043 Chr.19: 7847774-7847774 on Build GRCh38
rs371567316 Chr.19: 7847744-7847744 on Build GRCh38
rs375712197 Chr.19: 7847776-7847776 on Build GRCh38
rs778537610 Chr.19: 7847773-7847773 on Build GRCh38
rs757837977 Chr.19: 7847896-7847896 on Build GRCh38
rs146677119 Chr.19: 7847798-7847798 on Build GRCh38
rs768297573 Chr.19: 7847781-7847781 on Build GRCh38
rs367839369 Chr.19: 7847879-7847879 on Build GRCh38
rs193137808 Chr.19: 7847873-7847873 on Build GRCh38
rs761718634 Chr.19: 7847815-7847815 on Build GRCh38
rs752791877 Chr.19: 7847874-7847874 on Build GRCh38
rs766398564 Chr.19: 7847721-7847721 on Build GRCh38
rs753652084 Chr.19: 7847715-7847715 on Build GRCh38

View Full Product Details