Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 78940931-78941170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
UniGene
Hs.656212
Gene Chromosome Location
Chr.6: 78867486-78979411 on Build GRCh38
Transcripts
NM_017934.5 XM_005248729.4 XM_011535918.2 XM_017010989.1 XM_017010990.1

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
UniGene
Hs.511817
Gene Chromosome Location
Chr.6: 78934419-79078294 on Build GRCh38
Transcripts
NM_017934.5 XM_005248729.4 XM_011535918.2 XM_017010989.1 XM_017010990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426335 c.5052T>C p.D1684D Chr.6: 78941107-78941107 on Build GRCh38
743058 c.5046C>G p.I1682M Chr.6: 78941113-78941113 on Build GRCh38
246568 c.5054A>C p.E1685A Chr.6: 78941105-78941105 on Build GRCh38
4996758 c.5066C>G p.S1689C Chr.6: 78941093-78941093 on Build GRCh38
1446044 c.4991A>C p.K1664T Chr.6: 78941168-78941168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372154191 Chr.6: 78941047-78941047 on Build GRCh38
rs368832425 Chr.6: 78941023-78941023 on Build GRCh38
rs747724327 Chr.6: 78941098-78941098 on Build GRCh38
rs760828705 Chr.6: 78941056-78941056 on Build GRCh38
rs902930089 Chr.6: 78941094-78941094 on Build GRCh38
rs759804294 Chr.6: 78941124-78941124 on Build GRCh38
rs188746047 Chr.6: 78941128-78941128 on Build GRCh38
rs952159713 Chr.6: 78940983-78940983 on Build GRCh38
rs774746332 Chr.6: 78941027-78941027 on Build GRCh38
rs368703514 Chr.6: 78941139-78941139 on Build GRCh38
rs376750610 Chr.6: 78941081-78941081 on Build GRCh38
rs571545144 Chr.6: 78941086-78941086 on Build GRCh38
rs930842042 Chr.6: 78940966-78940966 on Build GRCh38
rs769265811 Chr.6: 78941108-78941108 on Build GRCh38
rs777455779 Chr.6: 78941116-78941116 on Build GRCh38
rs528882994 Chr.6: 78940972-78940972 on Build GRCh38
rs780978664 Chr.6: 78941096-78941096 on Build GRCh38
rs1011414294 Chr.6: 78941076-78941076 on Build GRCh38
rs549020329 Chr.6: 78940987-78940987 on Build GRCh38
rs774747260 Chr.6: 78941033-78941033 on Build GRCh38
rs768081139 Chr.6: 78940977-78940977 on Build GRCh38
rs551541016 Chr.6: 78941080-78941080 on Build GRCh38
rs754888929 Chr.6: 78941089-78941089 on Build GRCh38
rs771233799 Chr.6: 78941000-78941000 on Build GRCh38
rs748893859 Chr.6: 78941117-78941117 on Build GRCh38
rs34463462 Chr.6: 78940993-78940993 on Build GRCh38
rs771234803 Chr.6: 78941118-78941118 on Build GRCh38
rs375999660 Chr.6: 78940968-78940968 on Build GRCh38
rs565631371 Chr.6: 78941005-78941005 on Build GRCh38
rs754089631 Chr.6: 78941073-78941073 on Build GRCh38
rs202143050 Chr.6: 78941062-78941062 on Build GRCh38
rs776106601 Chr.6: 78941130-78941130 on Build GRCh38
rs768951489 Chr.6: 78940967-78940967 on Build GRCh38
rs760018505 Chr.6: 78941031-78941031 on Build GRCh38
rs774717285 Chr.6: 78941120-78941120 on Build GRCh38
rs373941762 Chr.6: 78940979-78940979 on Build GRCh38

View Full Product Details