Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 61753557-61753816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A2
Entrez Gene ID
338811
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Gene Aliases
TAFA-2, TAFA2
UniGene
Hs.269745
Gene Chromosome Location
Chr.12: 61708248-62192839 on Build GRCh38
Transcripts
NM_178539.4 XM_006719370.2 XM_011538272.1 XM_011538274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942335 c.344G>T p.G115V Chr.12: 61753662-61753662 on Build GRCh38
4756334 c.309G>C p.E103D Chr.12: 61753697-61753697 on Build GRCh38
468742 c.260-2A>T p.? Chr.12: 61753748-61753748 on Build GRCh38
942336 c.334G>A p.D112N Chr.12: 61753672-61753672 on Build GRCh38
5648697 c.337C>A p.R113R Chr.12: 61753669-61753669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201782050 Chr.12: 61753590-61753590 on Build GRCh38
rs200506288 Chr.12: 61753749-61753749 on Build GRCh38
rs772041207 Chr.12: 61753625-61753625 on Build GRCh38
rs761949624 Chr.12: 61753674-61753674 on Build GRCh38
rs112322074 Chr.12: 61753630-61753630 on Build GRCh38
rs944576878 Chr.12: 61753784-61753784 on Build GRCh38
rs868227999 Chr.12: 61753699-61753699 on Build GRCh38
rs765720603 Chr.12: 61753681-61753681 on Build GRCh38
rs765107842 Chr.12: 61753602-61753602 on Build GRCh38
rs756881941 Chr.12: 61753776-61753776 on Build GRCh38
rs200217706 Chr.12: 61753762-61753762 on Build GRCh38
rs1014004712 Chr.12: 61753625-61753626 on Build GRCh38
rs759536403 Chr.12: 61753758-61753758 on Build GRCh38
rs910367093 Chr.12: 61753774-61753774 on Build GRCh38
rs139053149 Chr.12: 61753671-61753671 on Build GRCh38
rs771719355 Chr.12: 61753757-61753758 on Build GRCh38
rs763205251 Chr.12: 61753740-61753740 on Build GRCh38
rs553294739 Chr.12: 61753750-61753750 on Build GRCh38
rs1025149540 Chr.12: 61753768-61753768 on Build GRCh38
rs149900113 Chr.12: 61753706-61753706 on Build GRCh38
rs569347374 Chr.12: 61753668-61753668 on Build GRCh38
rs575172184 Chr.12: 61753776-61753777 on Build GRCh38
rs763443717 Chr.12: 61753766-61753766 on Build GRCh38
rs566819525 Chr.12: 61753759-61753759 on Build GRCh38
rs770994630 Chr.12: 61753587-61753587 on Build GRCh38
rs35877412 Chr.12: 61753708-61753709 on Build GRCh38
rs775544130 Chr.12: 61753634-61753634 on Build GRCh38
rs551179878 Chr.12: 61753594-61753594 on Build GRCh38
rs899848659 Chr.12: 61753649-61753649 on Build GRCh38
rs570984869 Chr.12: 61753741-61753741 on Build GRCh38
rs772705632 Chr.12: 61753687-61753687 on Build GRCh38
rs776778644 Chr.12: 61753673-61753673 on Build GRCh38

View Full Product Details