Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9093541-9093810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1M1
Entrez Gene ID
125963
Gene Name
olfactory receptor family 1 subfamily M member 1
Gene Aliases
OR19-5, OR19-6
UniGene
Hs.553577
Gene Chromosome Location
Chr.19: 9093245-9094186 on Build GRCh38
Transcripts
NM_001004456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3701596 c.304T>C p.Y102H Chr.19: 9093548-9093548 on Build GRCh38
715110 c.439G>A p.A147T Chr.19: 9093683-9093683 on Build GRCh38
1494508 c.374C>T p.A125V Chr.19: 9093618-9093618 on Build GRCh38
7425525 c.323G>A p.G108D Chr.19: 9093567-9093567 on Build GRCh38
3836341 c.451T>A p.F151I Chr.19: 9093695-9093695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768588927 Chr.19: 9093590-9093590 on Build GRCh38
rs1026943324 Chr.19: 9093576-9093576 on Build GRCh38
rs747945091 Chr.19: 9093651-9093651 on Build GRCh38
rs778457151 Chr.19: 9093603-9093603 on Build GRCh38
rs747019370 Chr.19: 9093589-9093589 on Build GRCh38
rs923946328 Chr.19: 9093781-9093781 on Build GRCh38
rs371177684 Chr.19: 9093598-9093598 on Build GRCh38
rs753652444 Chr.19: 9093749-9093749 on Build GRCh38
rs199574381 Chr.19: 9093780-9093780 on Build GRCh38
rs375321461 Chr.19: 9093727-9093727 on Build GRCh38
rs368972960 Chr.19: 9093764-9093764 on Build GRCh38
rs754863155 Chr.19: 9093641-9093641 on Build GRCh38
rs752634717 Chr.19: 9093693-9093693 on Build GRCh38
rs185062607 Chr.19: 9093608-9093608 on Build GRCh38
rs777012159 Chr.19: 9093613-9093613 on Build GRCh38
rs151205505 Chr.19: 9093581-9093581 on Build GRCh38
rs773988415 Chr.19: 9093771-9093771 on Build GRCh38
rs368197862 Chr.19: 9093680-9093680 on Build GRCh38
rs376000674 Chr.19: 9093604-9093604 on Build GRCh38
rs572891081 Chr.19: 9093595-9093595 on Build GRCh38
rs769252571 Chr.19: 9093668-9093668 on Build GRCh38
rs746512574 Chr.19: 9093694-9093694 on Build GRCh38
rs551740717 Chr.19: 9093721-9093721 on Build GRCh38
rs770163695 Chr.19: 9093762-9093762 on Build GRCh38
rs757891606 Chr.19: 9093580-9093580 on Build GRCh38
rs770870710 Chr.19: 9093730-9093730 on Build GRCh38
rs757086989 Chr.19: 9093709-9093712 on Build GRCh38
rs188117149 Chr.19: 9093718-9093718 on Build GRCh38
rs759033050 Chr.19: 9093737-9093737 on Build GRCh38
rs781358922 Chr.19: 9093775-9093777 on Build GRCh38
rs747061382 Chr.19: 9093707-9093707 on Build GRCh38
rs997630611 Chr.19: 9093695-9093695 on Build GRCh38
rs556516727 Chr.19: 9093568-9093568 on Build GRCh38
rs749116821 Chr.19: 9093696-9093696 on Build GRCh38
rs112314731 Chr.19: 9093789-9093789 on Build GRCh38
rs774040929 Chr.19: 9093733-9093733 on Build GRCh38
rs79821860 Chr.19: 9093743-9093743 on Build GRCh38
rs755237502 Chr.19: 9093757-9093757 on Build GRCh38
rs375660862 Chr.19: 9093695-9093695 on Build GRCh38
rs751892302 Chr.19: 9093753-9093753 on Build GRCh38
rs968175675 Chr.19: 9093605-9093605 on Build GRCh38
rs145395343 Chr.19: 9093738-9093738 on Build GRCh38
rs372082145 Chr.19: 9093683-9093683 on Build GRCh38
rs765812666 Chr.19: 9093640-9093640 on Build GRCh38
rs4804097 Chr.19: 9093631-9093631 on Build GRCh38
rs372392767 Chr.19: 9093770-9093770 on Build GRCh38
rs201423732 Chr.19: 9093650-9093650 on Build GRCh38
rs199618873 Chr.19: 9093717-9093717 on Build GRCh38
rs139156930 Chr.19: 9093609-9093609 on Build GRCh38
rs201410610 Chr.19: 9093686-9093686 on Build GRCh38
rs779190474 Chr.19: 9093655-9093655 on Build GRCh38
rs201998994 Chr.19: 9093571-9093571 on Build GRCh38
rs755911679 Chr.19: 9093653-9093653 on Build GRCh38
rs760479348 Chr.19: 9093742-9093742 on Build GRCh38
rs756432945 Chr.19: 9093599-9093599 on Build GRCh38
rs747114660 Chr.19: 9093662-9093662 on Build GRCh38
rs369713413 Chr.19: 9093697-9093697 on Build GRCh38
rs892952412 Chr.19: 9093677-9093677 on Build GRCh38
rs199757859 Chr.19: 9093669-9093669 on Build GRCh38
rs772344043 Chr.19: 9093606-9093606 on Build GRCh38
rs868709297 Chr.19: 9093706-9093706 on Build GRCh38
rs147905359 Chr.19: 9093659-9093659 on Build GRCh38
rs200806246 Chr.19: 9093756-9093756 on Build GRCh38
rs773402821 Chr.19: 9093722-9093722 on Build GRCh38
rs141423466 Chr.19: 9093700-9093700 on Build GRCh38
rs750096175 Chr.19: 9093637-9093637 on Build GRCh38
rs148262558 Chr.19: 9093614-9093614 on Build GRCh38
rs769583135 Chr.19: 9093759-9093759 on Build GRCh38
rs143401934 Chr.19: 9093751-9093751 on Build GRCh38
rs148343734 Chr.19: 9093752-9093752 on Build GRCh38
rs773634110 Chr.19: 9093675-9093675 on Build GRCh38
rs772434948 Chr.19: 9093660-9093660 on Build GRCh38
rs567733875 Chr.19: 9093763-9093763 on Build GRCh38
rs748284523 Chr.19: 9093760-9093760 on Build GRCh38
rs771778997 Chr.19: 9093782-9093782 on Build GRCh38
rs780910231 Chr.19: 9093594-9093594 on Build GRCh38
rs115262881 Chr.19: 9093572-9093572 on Build GRCh38
rs7259007 Chr.19: 9093682-9093682 on Build GRCh38
rs748541045 Chr.19: 9093720-9093720 on Build GRCh38

View Full Product Details