Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57026190-57026415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
UniGene
Hs.528836
Gene Chromosome Location
Chr.16: 56989547-57083524 on Build GRCh38
Transcripts
NM_032206.4 XM_005256193.2 XM_005256194.3 XM_005256195.2 XM_005256196.2 XM_005256197.2 XM_005256199.3 XM_005256201.3 XM_006721297.2 XM_006721298.2 XM_006721300.2 XM_011523373.2 XM_011523375.2 XM_017023766.1 XM_017023767.1 XM_017023768.1 XM_017023769.1 XM_017023770.1 XM_017023771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5680023 c.1309C>G p.Q437E Chr.16: 57026252-57026252 on Build GRCh38
5886930 c.1261C>T p.P421S Chr.16: 57026204-57026204 on Build GRCh38
4061378 c.1394T>C p.L465P Chr.16: 57026337-57026337 on Build GRCh38
5499914 c.1366T>G p.S456A Chr.16: 57026309-57026309 on Build GRCh38
4061377 c.1321C>T p.Q441* Chr.16: 57026264-57026264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753432434 Chr.16: 57026287-57026287 on Build GRCh38
rs376911783 Chr.16: 57026246-57026246 on Build GRCh38
rs770326888 Chr.16: 57026259-57026259 on Build GRCh38
rs1038993953 Chr.16: 57026321-57026321 on Build GRCh38
rs764789454 Chr.16: 57026275-57026281 on Build GRCh38
rs763365261 Chr.16: 57026273-57026273 on Build GRCh38
rs900622944 Chr.16: 57026333-57026333 on Build GRCh38
rs748990366 Chr.16: 57026291-57026291 on Build GRCh38
rs267604583 Chr.16: 57026310-57026310 on Build GRCh38
rs762883583 Chr.16: 57026322-57026323 on Build GRCh38
rs373548722 Chr.16: 57026320-57026320 on Build GRCh38
rs566557427 Chr.16: 57026245-57026245 on Build GRCh38
rs144419797 Chr.16: 57026308-57026308 on Build GRCh38
rs545626603 Chr.16: 57026370-57026370 on Build GRCh38
rs556886442 Chr.16: 57026339-57026339 on Build GRCh38
rs142400015 Chr.16: 57026360-57026360 on Build GRCh38
rs775917017 Chr.16: 57026336-57026336 on Build GRCh38
rs866303781 Chr.16: 57026210-57026210 on Build GRCh38
rs267604584 Chr.16: 57026311-57026311 on Build GRCh38
rs371278281 Chr.16: 57026262-57026262 on Build GRCh38
rs139094752 Chr.16: 57026352-57026352 on Build GRCh38
rs950510637 Chr.16: 57026384-57026384 on Build GRCh38
rs770877382 Chr.16: 57026228-57026228 on Build GRCh38
rs149735722 Chr.16: 57026224-57026224 on Build GRCh38
rs994922009 Chr.16: 57026359-57026359 on Build GRCh38
rs763777727 Chr.16: 57026378-57026378 on Build GRCh38
rs752699384 Chr.16: 57026289-57026289 on Build GRCh38
rs878972384 Chr.16: 57026307-57026307 on Build GRCh38
rs760218012 Chr.16: 57026340-57026340 on Build GRCh38
rs143613942 Chr.16: 57026209-57026209 on Build GRCh38
rs202133400 Chr.16: 57026276-57026276 on Build GRCh38
rs150148526 Chr.16: 57026212-57026212 on Build GRCh38
rs926111764 Chr.16: 57026285-57026285 on Build GRCh38
rs772651939 Chr.16: 57026283-57026284 on Build GRCh38
rs764453977 Chr.16: 57026275-57026275 on Build GRCh38
rs752644497 Chr.16: 57026364-57026364 on Build GRCh38
rs765755141 Chr.16: 57026277-57026277 on Build GRCh38
rs778427595 Chr.16: 57026213-57026213 on Build GRCh38
rs34763272 Chr.16: 57026343-57026344 on Build GRCh38
rs551852285 Chr.16: 57026232-57026232 on Build GRCh38
rs749799828 Chr.16: 57026338-57026338 on Build GRCh38
rs9938543 Chr.16: 57026301-57026301 on Build GRCh38
rs770271872 Chr.16: 57026332-57026332 on Build GRCh38
rs201280147 Chr.16: 57026288-57026288 on Build GRCh38
rs200312421 Chr.16: 57026343-57026343 on Build GRCh38
rs777745465 Chr.16: 57026290-57026290 on Build GRCh38

View Full Product Details