Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 72513023-72513296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6B
Entrez Gene ID
23233
Gene Name
exocyst complex component 6B
Gene Aliases
SEC15B, SEC15L2
UniGene
Hs.303454
Gene Chromosome Location
Chr.2: 72175984-72826042 on Build GRCh38
Transcripts
NM_001321729.1 NM_001321730.1 NM_001321731.1 NM_001321733.1 NM_001321734.1 NM_015189.2 NR_135773.1 NR_135774.1 XM_005264224.1 XM_011532711.2 XM_011532712.2 XM_017003641.1 XM_017003642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5397350 c.1147G>T p.A383S Chr.2: 72513152-72513152 on Build GRCh38
4624857 c.1094T>C p.V365A Chr.2: 72513205-72513205 on Build GRCh38
359458 c.1149A>T p.A383A Chr.2: 72513150-72513150 on Build GRCh38
1251596 c.1119G>T p.L373L Chr.2: 72513180-72513180 on Build GRCh38
1022718 c.1095A>T p.V365V Chr.2: 72513204-72513204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76275893 Chr.2: 72513174-72513174 on Build GRCh38
rs143178994 Chr.2: 72513097-72513097 on Build GRCh38
rs753817492 Chr.2: 72513153-72513153 on Build GRCh38
rs751729296 Chr.2: 72513145-72513145 on Build GRCh38
rs894265023 Chr.2: 72513058-72513058 on Build GRCh38
rs993101724 Chr.2: 72513216-72513216 on Build GRCh38
rs746477963 Chr.2: 72513222-72513222 on Build GRCh38
rs770203010 Chr.2: 72513109-72513109 on Build GRCh38
rs1026581200 Chr.2: 72513242-72513242 on Build GRCh38
rs746457066 Chr.2: 72513266-72513266 on Build GRCh38
rs764248141 Chr.2: 72513136-72513136 on Build GRCh38
rs766170220 Chr.2: 72513152-72513152 on Build GRCh38
rs1039920831 Chr.2: 72513059-72513059 on Build GRCh38
rs199672443 Chr.2: 72513143-72513143 on Build GRCh38
rs775572928 Chr.2: 72513117-72513117 on Build GRCh38
rs773843607 Chr.2: 72513095-72513098 on Build GRCh38
rs777220976 Chr.2: 72513100-72513100 on Build GRCh38
rs542067463 Chr.2: 72513209-72513209 on Build GRCh38
rs752321034 Chr.2: 72513186-72513186 on Build GRCh38
rs781488675 Chr.2: 72513225-72513225 on Build GRCh38
rs758083040 Chr.2: 72513189-72513189 on Build GRCh38
rs759765454 Chr.2: 72513105-72513105 on Build GRCh38
rs762014732 Chr.2: 72513147-72513147 on Build GRCh38
rs770445435 Chr.2: 72513224-72513224 on Build GRCh38
rs1010005137 Chr.2: 72513248-72513248 on Build GRCh38
rs368207726 Chr.2: 72513118-72513118 on Build GRCh38
rs35801064 Chr.2: 72513114-72513115 on Build GRCh38
rs777311063 Chr.2: 72513087-72513087 on Build GRCh38
rs747479638 Chr.2: 72513089-72513089 on Build GRCh38
rs754722759 Chr.2: 72513172-72513172 on Build GRCh38
rs553890023 Chr.2: 72513061-72513061 on Build GRCh38
rs752886733 Chr.2: 72513133-72513133 on Build GRCh38
rs868200677 Chr.2: 72513155-72513155 on Build GRCh38
rs770044685 Chr.2: 72513268-72513268 on Build GRCh38

View Full Product Details