Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74740068-74740329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA2022
Entrez Gene ID
340533
Gene Name
KIAA2022
Gene Aliases
MRX98, XPN
UniGene
Hs.124128
Gene Chromosome Location
Chr.X: 74732856-74925452 on Build GRCh38
Transcripts
NM_001008537.2

SNP Detail

SNP ID Chromosome Location
rs756589710 Chr.X: 74740299-74740299 on Build GRCh38
rs776659282 Chr.X: 74740112-74740112 on Build GRCh38
rs148446893 Chr.X: 74740108-74740108 on Build GRCh38
rs868554078 Chr.X: 74740297-74740297 on Build GRCh38
rs371190782 Chr.X: 74740135-74740135 on Build GRCh38
rs764469950 Chr.X: 74740166-74740166 on Build GRCh38
rs775723064 Chr.X: 74740243-74740243 on Build GRCh38
rs771736722 Chr.X: 74740220-74740220 on Build GRCh38
rs750798131 Chr.X: 74740296-74740296 on Build GRCh38
rs770781682 Chr.X: 74740111-74740111 on Build GRCh38
rs759758425 Chr.X: 74740127-74740127 on Build GRCh38
rs769979162 Chr.X: 74740237-74740237 on Build GRCh38
rs200322426 Chr.X: 74740290-74740290 on Build GRCh38
rs781623230 Chr.X: 74740189-74740189 on Build GRCh38
rs141163613 Chr.X: 74740231-74740231 on Build GRCh38
rs151004644 Chr.X: 74740151-74740151 on Build GRCh38
rs764248445 Chr.X: 74740263-74740263 on Build GRCh38
rs142635989 Chr.X: 74740143-74740143 on Build GRCh38
rs1057518728 Chr.X: 74740308-74740309 on Build GRCh38
rs1041986802 Chr.X: 74740123-74740123 on Build GRCh38
rs760172736 Chr.X: 74740180-74740180 on Build GRCh38
rs778760434 Chr.X: 74740211-74740211 on Build GRCh38
rs754667761 Chr.X: 74740198-74740198 on Build GRCh38
rs370429318 Chr.X: 74740232-74740232 on Build GRCh38
rs752130414 Chr.X: 74740173-74740173 on Build GRCh38
rs975233116 Chr.X: 74740093-74740093 on Build GRCh38
rs762973583 Chr.X: 74740250-74740250 on Build GRCh38
rs1057098571 Chr.X: 74740158-74740158 on Build GRCh38
rs373663454 Chr.X: 74740174-74740174 on Build GRCh38
rs865796472 Chr.X: 74740182-74740182 on Build GRCh38
rs774654591 Chr.X: 74740268-74740268 on Build GRCh38
rs750888177 Chr.X: 74740190-74740190 on Build GRCh38
rs764940962 Chr.X: 74740300-74740300 on Build GRCh38
rs752417405 Chr.X: 74740304-74740304 on Build GRCh38
rs143577015 Chr.X: 74740311-74740311 on Build GRCh38
rs750153892 Chr.X: 74740289-74740289 on Build GRCh38
rs199998866 Chr.X: 74740148-74740148 on Build GRCh38
rs954974268 Chr.X: 74740267-74740267 on Build GRCh38
rs763067676 Chr.X: 74740152-74740152 on Build GRCh38
rs776481180 Chr.X: 74740235-74740235 on Build GRCh38
rs946737858 Chr.X: 74740212-74740212 on Build GRCh38
rs200805356 Chr.X: 74740219-74740219 on Build GRCh38
rs757785426 Chr.X: 74740306-74740306 on Build GRCh38
rs745720102 Chr.X: 74740236-74740236 on Build GRCh38

View Full Product Details