Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 25395840-25396101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR158
Entrez Gene ID
57512
Gene Name
G protein-coupled receptor 158
Gene Aliases
-
UniGene
Hs.499108
Gene Chromosome Location
Chr.10: 25174722-25602229 on Build GRCh38
Transcripts
NM_020752.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161350 c.1039C>T p.L347F Chr.10: 25395941-25395941 on Build GRCh38
6480585 c.1029A>C p.L343L Chr.10: 25395931-25395931 on Build GRCh38
5603675 c.1103A>G p.N368S Chr.10: 25396005-25396005 on Build GRCh38
683605 c.1081C>A p.P361T Chr.10: 25395983-25395983 on Build GRCh38
3867044 c.1098G>A p.V366V Chr.10: 25396000-25396000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369165750 Chr.10: 25395938-25395938 on Build GRCh38
rs761783401 Chr.10: 25395947-25395947 on Build GRCh38
rs746213398 Chr.10: 25395886-25395886 on Build GRCh38
rs536930029 Chr.10: 25395926-25395926 on Build GRCh38
rs149833592 Chr.10: 25395981-25395981 on Build GRCh38
rs762550852 Chr.10: 25395925-25395925 on Build GRCh38
rs545538384 Chr.10: 25395985-25395985 on Build GRCh38
rs753885823 Chr.10: 25396042-25396042 on Build GRCh38
rs372485855 Chr.10: 25396033-25396033 on Build GRCh38
rs772733643 Chr.10: 25395921-25395921 on Build GRCh38
rs759230110 Chr.10: 25396013-25396013 on Build GRCh38
rs772263062 Chr.10: 25395984-25395984 on Build GRCh38
rs906597881 Chr.10: 25395872-25395872 on Build GRCh38
rs772545248 Chr.10: 25395893-25395893 on Build GRCh38
rs775860988 Chr.10: 25395895-25395895 on Build GRCh38
rs767413538 Chr.10: 25395937-25395937 on Build GRCh38
rs375234517 Chr.10: 25395892-25395892 on Build GRCh38
rs767158539 Chr.10: 25396015-25396015 on Build GRCh38
rs757408413 Chr.10: 25396059-25396059 on Build GRCh38
rs773988207 Chr.10: 25396011-25396011 on Build GRCh38
rs756143078 Chr.10: 25395868-25395868 on Build GRCh38
rs141199449 Chr.10: 25396010-25396010 on Build GRCh38
rs764102789 Chr.10: 25396039-25396039 on Build GRCh38
rs774755368 Chr.10: 25395885-25395885 on Build GRCh38
rs749462111 Chr.10: 25395875-25395875 on Build GRCh38
rs139869897 Chr.10: 25395942-25395942 on Build GRCh38
rs760428535 Chr.10: 25396037-25396037 on Build GRCh38
rs753940988 Chr.10: 25395955-25395955 on Build GRCh38
rs1005726641 Chr.10: 25396005-25396005 on Build GRCh38
rs764596692 Chr.10: 25395903-25395903 on Build GRCh38
rs762424522 Chr.10: 25396004-25396004 on Build GRCh38
rs200721685 Chr.10: 25395920-25395920 on Build GRCh38
rs757076266 Chr.10: 25396035-25396036 on Build GRCh38
rs777160161 Chr.10: 25395994-25395994 on Build GRCh38
rs551640586 Chr.10: 25396060-25396060 on Build GRCh38
rs769239585 Chr.10: 25395987-25395987 on Build GRCh38
rs777911788 Chr.10: 25395869-25395869 on Build GRCh38
rs181041655 Chr.10: 25395969-25395969 on Build GRCh38
rs771311638 Chr.10: 25395884-25395884 on Build GRCh38
rs373396589 Chr.10: 25395928-25395928 on Build GRCh38
rs145500194 Chr.10: 25395983-25395983 on Build GRCh38
rs376414773 Chr.10: 25395929-25395929 on Build GRCh38

View Full Product Details