Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 8734634-8734875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIDINS220
Entrez Gene ID
57498
Gene Name
kinase D-interacting substrate 220kDa
Gene Aliases
ARMS
UniGene
Hs.9873
Gene Chromosome Location
Chr.2: 8728857-8837625 on Build GRCh38
Transcripts
NM_020738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1483337 c.3796T>C p.W1266R Chr.2: 8734675-8734675 on Build GRCh38
4822418 c.3777G>A p.M1259I Chr.2: 8734694-8734694 on Build GRCh38
1471357 c.3787T>G p.F1263V Chr.2: 8734684-8734684 on Build GRCh38
6370158 c.3753C>A p.N1251K Chr.2: 8734718-8734718 on Build GRCh38
7110533 c.3772G>C p.E1258Q Chr.2: 8734699-8734699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769995477 Chr.2: 8734786-8734786 on Build GRCh38
rs763222825 Chr.2: 8734711-8734711 on Build GRCh38
rs779667005 Chr.2: 8734787-8734787 on Build GRCh38
rs748597203 Chr.2: 8734692-8734692 on Build GRCh38
rs773517730 Chr.2: 8734710-8734710 on Build GRCh38
rs922595537 Chr.2: 8734827-8734827 on Build GRCh38
rs759687436 Chr.2: 8734738-8734738 on Build GRCh38
rs959541105 Chr.2: 8734735-8734735 on Build GRCh38
rs753698931 Chr.2: 8734750-8734750 on Build GRCh38
rs767889901 Chr.2: 8734696-8734697 on Build GRCh38
rs984192913 Chr.2: 8734764-8734764 on Build GRCh38
rs764902447 Chr.2: 8734774-8734774 on Build GRCh38
rs750095410 Chr.2: 8734780-8734780 on Build GRCh38
rs764170262 Chr.2: 8734747-8734747 on Build GRCh38
rs776992785 Chr.2: 8734671-8734671 on Build GRCh38
rs199769091 Chr.2: 8734753-8734753 on Build GRCh38
rs752924094 Chr.2: 8734760-8734760 on Build GRCh38
rs985791881 Chr.2: 8734841-8734841 on Build GRCh38
rs984352671 Chr.2: 8734767-8734767 on Build GRCh38
rs751136124 Chr.2: 8734794-8734794 on Build GRCh38
rs770026451 Chr.2: 8734697-8734697 on Build GRCh38
rs757098860 Chr.2: 8734768-8734768 on Build GRCh38
rs766567862 Chr.2: 8734724-8734724 on Build GRCh38
rs143217814 Chr.2: 8734781-8734781 on Build GRCh38
rs377648666 Chr.2: 8734717-8734717 on Build GRCh38
rs955291990 Chr.2: 8734831-8734831 on Build GRCh38
rs972651134 Chr.2: 8734848-8734848 on Build GRCh38
rs61742970 Chr.2: 8734737-8734737 on Build GRCh38

View Full Product Details