Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79411381-79411616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC1
Entrez Gene ID
4772
Gene Name
nuclear factor of activated T-cells 1
Gene Aliases
NF-ATC, NF-ATc1.2, NFAT2, NFATc
UniGene
Hs.534074
Gene Chromosome Location
Chr.18: 79395772-79529323 on Build GRCh38
Transcripts
NM_001278669.1 NM_001278670.1 NM_001278672.1 NM_001278673.1 NM_001278675.1 NM_006162.4 NM_172387.2 NM_172388.2 NM_172389.2 NM_172390.2 XM_017025783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364546 c.1151C>T p.A384V Chr.18: 79411465-79411465 on Build GRCh38
5667519 c.1189G>T p.A397S Chr.18: 79411464-79411464 on Build GRCh38
3527800 c.1171C>T p.P391S Chr.18: 79411485-79411485 on Build GRCh38
3147392 c.1178C>T p.S393F Chr.18: 79411492-79411492 on Build GRCh38
5626084 c.1178C>T p.P393L Chr.18: 79411453-79411453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766116077 Chr.18: 79411517-79411517 on Build GRCh38
rs745932419 Chr.18: 79411514-79411515 on Build GRCh38
rs901204214 Chr.18: 79411596-79411596 on Build GRCh38
rs758837258 Chr.18: 79411519-79411519 on Build GRCh38
rs760274443 Chr.18: 79411549-79411549 on Build GRCh38
rs940347722 Chr.18: 79411567-79411567 on Build GRCh38
rs745936820 Chr.18: 79411528-79411528 on Build GRCh38
rs201530582 Chr.18: 79411465-79411465 on Build GRCh38
rs866281675 Chr.18: 79411450-79411450 on Build GRCh38
rs999505939 Chr.18: 79411597-79411597 on Build GRCh38
rs372345100 Chr.18: 79411447-79411447 on Build GRCh38
rs775236204 Chr.18: 79411453-79411453 on Build GRCh38
rs368527191 Chr.18: 79411445-79411445 on Build GRCh38
rs762946877 Chr.18: 79411461-79411461 on Build GRCh38
rs542179762 Chr.18: 79411490-79411490 on Build GRCh38
rs747406753 Chr.18: 79411483-79411483 on Build GRCh38
rs781111342 Chr.18: 79411486-79411486 on Build GRCh38
rs538775407 Chr.18: 79411478-79411478 on Build GRCh38
rs748153451 Chr.18: 79411545-79411545 on Build GRCh38
rs760468564 Chr.18: 79411516-79411516 on Build GRCh38
rs773991329 Chr.18: 79411540-79411540 on Build GRCh38
rs916823045 Chr.18: 79411532-79411532 on Build GRCh38
rs920333860 Chr.18: 79411571-79411571 on Build GRCh38
rs565127643 Chr.18: 79411534-79411534 on Build GRCh38
rs769809992 Chr.18: 79411529-79411529 on Build GRCh38
rs768522089 Chr.18: 79411535-79411535 on Build GRCh38
rs768273257 Chr.18: 79411508-79411508 on Build GRCh38
rs888995264 Chr.18: 79411515-79411519 on Build GRCh38
rs757674978 Chr.18: 79411525-79411525 on Build GRCh38
rs927655705 Chr.18: 79411561-79411561 on Build GRCh38
rs1008858301 Chr.18: 79411583-79411583 on Build GRCh38
rs199736877 Chr.18: 79411438-79411438 on Build GRCh38
rs781348090 Chr.18: 79411510-79411514 on Build GRCh38
rs781401849 Chr.18: 79411527-79411527 on Build GRCh38
rs774380800 Chr.18: 79411510-79411510 on Build GRCh38
rs899202080 Chr.18: 79411407-79411407 on Build GRCh38
rs368411554 Chr.18: 79411514-79411514 on Build GRCh38
rs766901147 Chr.18: 79411466-79411466 on Build GRCh38
rs1050235585 Chr.18: 79411578-79411587 on Build GRCh38
rs540117854 Chr.18: 79411523-79411523 on Build GRCh38
rs767099121 Chr.18: 79411513-79411513 on Build GRCh38
rs780292393 Chr.18: 79411520-79411521 on Build GRCh38
rs930474982 Chr.18: 79411574-79411574 on Build GRCh38
rs768947037 Chr.18: 79411527-79411536 on Build GRCh38
rs199859549 Chr.18: 79411469-79411469 on Build GRCh38
rs746130678 Chr.18: 79411489-79411489 on Build GRCh38
rs745449379 Chr.18: 79411404-79411404 on Build GRCh38
rs772912940 Chr.18: 79411547-79411547 on Build GRCh38
rs770858652 Chr.18: 79411439-79411439 on Build GRCh38
rs776916137 Chr.18: 79411444-79411444 on Build GRCh38
rs532203776 Chr.18: 79411556-79411556 on Build GRCh38
rs1036533618 Chr.18: 79411570-79411570 on Build GRCh38
rs146992531 Chr.18: 79411416-79411416 on Build GRCh38
rs1040297045 Chr.18: 79411588-79411588 on Build GRCh38
rs764008108 Chr.18: 79411464-79411464 on Build GRCh38
rs755801441 Chr.18: 79411472-79411472 on Build GRCh38
rs762752510 Chr.18: 79411493-79411493 on Build GRCh38
rs888944399 Chr.18: 79411582-79411582 on Build GRCh38
rs200663312 Chr.18: 79411480-79411480 on Build GRCh38
rs765918984 Chr.18: 79411551-79411551 on Build GRCh38
rs771065897 Chr.18: 79411485-79411485 on Build GRCh38
rs775324687 Chr.18: 79411492-79411492 on Build GRCh38
rs113881412 Chr.18: 79411518-79411518 on Build GRCh38
rs755421228 Chr.18: 79411412-79411412 on Build GRCh38
rs200186658 Chr.18: 79411424-79411424 on Build GRCh38
rs200146253 Chr.18: 79411481-79411481 on Build GRCh38
rs201480996 Chr.18: 79411522-79411522 on Build GRCh38
rs1031073719 Chr.18: 79411526-79411526 on Build GRCh38
rs761603161 Chr.18: 79411511-79411511 on Build GRCh38
rs771897134 Chr.18: 79411546-79411546 on Build GRCh38
rs191625409 Chr.18: 79411415-79411415 on Build GRCh38
rs780262289 Chr.18: 79411530-79411530 on Build GRCh38
rs570950086 Chr.18: 79411476-79411476 on Build GRCh38
rs749431312 Chr.18: 79411526-79411527 on Build GRCh38

View Full Product Details