Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14965486-14965737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRIP1
Entrez Gene ID
8204
Gene Name
nuclear receptor interacting protein 1
Gene Aliases
RIP140
UniGene
Hs.155017
Gene Chromosome Location
Chr.21: 14961235-15065903 on Build GRCh38
Transcripts
NM_003489.3 XM_005261063.3 XM_005261065.3 XM_011529747.1 XM_011529748.2 XM_011529749.2 XM_011529751.2 XM_011529752.1 XM_017028473.1 XM_017028474.1 XM_017028475.1 XM_017028476.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3939467 c.2584T>C p.Y862H Chr.21: 14965609-14965609 on Build GRCh38
3939468 c.2546C>G p.S849* Chr.21: 14965647-14965647 on Build GRCh38
725494 c.2466G>A p.L822L Chr.21: 14965727-14965727 on Build GRCh38
7285774 c.2567C>A p.P856H Chr.21: 14965626-14965626 on Build GRCh38
5392714 c.2606C>T p.P869L Chr.21: 14965587-14965587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369884270 Chr.21: 14965639-14965639 on Build GRCh38
rs756506808 Chr.21: 14965572-14965572 on Build GRCh38
rs759612473 Chr.21: 14965564-14965564 on Build GRCh38
rs1028853539 Chr.21: 14965657-14965657 on Build GRCh38
rs779601974 Chr.21: 14965686-14965686 on Build GRCh38
rs756970047 Chr.21: 14965585-14965585 on Build GRCh38
rs554495028 Chr.21: 14965594-14965594 on Build GRCh38
rs371257897 Chr.21: 14965552-14965552 on Build GRCh38
rs376704589 Chr.21: 14965612-14965612 on Build GRCh38
rs750058770 Chr.21: 14965673-14965673 on Build GRCh38
rs778756254 Chr.21: 14965659-14965659 on Build GRCh38
rs556159450 Chr.21: 14965643-14965643 on Build GRCh38
rs1027440653 Chr.21: 14965558-14965558 on Build GRCh38
rs771165207 Chr.21: 14965635-14965635 on Build GRCh38
rs192032924 Chr.21: 14965631-14965631 on Build GRCh38
rs773489507 Chr.21: 14965545-14965545 on Build GRCh38
rs776416018 Chr.21: 14965628-14965628 on Build GRCh38
rs751624261 Chr.21: 14965557-14965557 on Build GRCh38
rs757643881 Chr.21: 14965652-14965652 on Build GRCh38
rs753219568 Chr.21: 14965569-14965569 on Build GRCh38
rs754939703 Chr.21: 14965704-14965704 on Build GRCh38
rs778666371 Chr.21: 14965586-14965586 on Build GRCh38
rs143359165 Chr.21: 14965642-14965642 on Build GRCh38
rs753205102 Chr.21: 14965641-14965641 on Build GRCh38
rs763176540 Chr.21: 14965546-14965546 on Build GRCh38
rs374602006 Chr.21: 14965650-14965650 on Build GRCh38
rs1001187599 Chr.21: 14965548-14965548 on Build GRCh38
rs778272175 Chr.21: 14965574-14965574 on Build GRCh38
rs761081090 Chr.21: 14965627-14965627 on Build GRCh38
rs976427561 Chr.21: 14965653-14965653 on Build GRCh38
rs779895179 Chr.21: 14965599-14965599 on Build GRCh38
rs768408644 Chr.21: 14965521-14965521 on Build GRCh38
rs745389893 Chr.21: 14965588-14965588 on Build GRCh38
rs368239131 Chr.21: 14965684-14965684 on Build GRCh38
rs774613316 Chr.21: 14965636-14965636 on Build GRCh38
rs746437857 Chr.21: 14965698-14965698 on Build GRCh38
rs767634577 Chr.21: 14965640-14965640 on Build GRCh38
rs754193620 Chr.21: 14965575-14965575 on Build GRCh38

View Full Product Details