Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50265203-50265470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L2HGDH
Entrez Gene ID
79944
Gene Name
L-2-hydroxyglutarate dehydrogenase
Gene Aliases
C14orf160, L2HGA
UniGene
Hs.256034
Gene Chromosome Location
Chr.14: 50242434-50312235 on Build GRCh38
Transcripts
NM_024884.2 XM_005268075.4 XM_011537166.2 XM_011537167.2 XM_011537168.2 XM_017021655.1 XM_017021656.1 XM_017021657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3814861 c.1196+1G>A p.? Chr.14: 50265357-50265357 on Build GRCh38
4458494 c.1087C>T p.Q363* Chr.14: 50265467-50265467 on Build GRCh38
3496169 c.1168C>T p.P390S Chr.14: 50265386-50265386 on Build GRCh38
3496170 c.1168C>T p.P390S Chr.14: 50265386-50265386 on Build GRCh38
3886073 c.1097C>T p.S366F Chr.14: 50265457-50265457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053062980 Chr.14: 50265436-50265436 on Build GRCh38
rs786200869 Chr.14: 50265439-50265439 on Build GRCh38
rs746576098 Chr.14: 50265274-50265274 on Build GRCh38
rs183258819 Chr.14: 50265341-50265341 on Build GRCh38
rs770155002 Chr.14: 50265370-50265370 on Build GRCh38
rs150157112 Chr.14: 50265377-50265377 on Build GRCh38
rs12432673 Chr.14: 50265324-50265324 on Build GRCh38
rs748551420 Chr.14: 50265364-50265364 on Build GRCh38
rs199809061 Chr.14: 50265328-50265328 on Build GRCh38
rs749644737 Chr.14: 50265375-50265375 on Build GRCh38
rs1045128582 Chr.14: 50265270-50265270 on Build GRCh38
rs529020131 Chr.14: 50265285-50265285 on Build GRCh38
rs773495683 Chr.14: 50265374-50265374 on Build GRCh38
rs925420882 Chr.14: 50265299-50265299 on Build GRCh38
rs750550838 Chr.14: 50265323-50265323 on Build GRCh38
rs925835899 Chr.14: 50265422-50265422 on Build GRCh38
rs141715606 Chr.14: 50265399-50265399 on Build GRCh38
rs528333358 Chr.14: 50265316-50265316 on Build GRCh38
rs775513079 Chr.14: 50265418-50265418 on Build GRCh38
rs141017067 Chr.14: 50265349-50265349 on Build GRCh38
rs372144735 Chr.14: 50265433-50265433 on Build GRCh38
rs373172891 Chr.14: 50265380-50265380 on Build GRCh38
rs750679503 Chr.14: 50265335-50265335 on Build GRCh38
rs146471399 Chr.14: 50265301-50265301 on Build GRCh38
rs948177479 Chr.14: 50265269-50265269 on Build GRCh38
rs747160991 Chr.14: 50265342-50265342 on Build GRCh38
rs376430390 Chr.14: 50265307-50265307 on Build GRCh38
rs892740235 Chr.14: 50265338-50265338 on Build GRCh38
rs879171135 Chr.14: 50265400-50265400 on Build GRCh38
rs917991536 Chr.14: 50265252-50265252 on Build GRCh38
rs768586031 Chr.14: 50265346-50265346 on Build GRCh38
rs201917411 Chr.14: 50265326-50265326 on Build GRCh38
rs375346810 Chr.14: 50265376-50265376 on Build GRCh38
rs760624214 Chr.14: 50265424-50265424 on Build GRCh38
rs758761286 Chr.14: 50265337-50265337 on Build GRCh38
rs370203574 Chr.14: 50265317-50265317 on Build GRCh38
rs368843696 Chr.14: 50265333-50265333 on Build GRCh38
rs764123304 Chr.14: 50265428-50265428 on Build GRCh38
rs753744256 Chr.14: 50265432-50265432 on Build GRCh38

View Full Product Details