Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74738549-74738800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYW3
Entrez Gene ID
127253
Gene Name
tRNA-yW synthesizing protein 3 homolog
Gene Aliases
C1orf171
UniGene
Hs.348411
Gene Chromosome Location
Chr.1: 74733143-74766677 on Build GRCh38
Transcripts
NM_001162916.1 NM_138467.2 NR_027962.1 XM_006710347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4738556 c.337C>T p.Q113* Chr.1: 74738771-74738771 on Build GRCh38
1344295 c.304C>T p.P102S Chr.1: 74738738-74738738 on Build GRCh38
682121 c.305C>T p.P102L Chr.1: 74738739-74738739 on Build GRCh38
3790257 c.283G>A p.D95N Chr.1: 74738717-74738717 on Build GRCh38
136142 c.263C>T p.A88V Chr.1: 74738697-74738697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775960089 Chr.1: 74738652-74738652 on Build GRCh38
rs372873769 Chr.1: 74738718-74738718 on Build GRCh38
rs796332443 Chr.1: 74738732-74738732 on Build GRCh38
rs111491709 Chr.1: 74738663-74738663 on Build GRCh38
rs758441583 Chr.1: 74738717-74738717 on Build GRCh38
rs757114839 Chr.1: 74738738-74738738 on Build GRCh38
rs183955945 Chr.1: 74738572-74738572 on Build GRCh38
rs867006317 Chr.1: 74738653-74738653 on Build GRCh38
rs745588555 Chr.1: 74738757-74738757 on Build GRCh38
rs531959275 Chr.1: 74738674-74738674 on Build GRCh38
rs768938416 Chr.1: 74738670-74738670 on Build GRCh38
rs752615719 Chr.1: 74738714-74738714 on Build GRCh38
rs760659488 Chr.1: 74738707-74738707 on Build GRCh38
rs572825166 Chr.1: 74738620-74738620 on Build GRCh38
rs969120656 Chr.1: 74738752-74738752 on Build GRCh38
rs1013589376 Chr.1: 74738616-74738616 on Build GRCh38
rs766580523 Chr.1: 74738711-74738711 on Build GRCh38
rs11538281 Chr.1: 74738763-74738763 on Build GRCh38
rs982516286 Chr.1: 74738637-74738637 on Build GRCh38
rs755560668 Chr.1: 74738762-74738762 on Build GRCh38
rs762290911 Chr.1: 74738676-74738676 on Build GRCh38
rs965403424 Chr.1: 74738611-74738611 on Build GRCh38
rs958954149 Chr.1: 74738579-74738579 on Build GRCh38
rs769357652 Chr.1: 74738642-74738642 on Build GRCh38
rs867439371 Chr.1: 74738654-74738654 on Build GRCh38
rs562435731 Chr.1: 74738744-74738744 on Build GRCh38
rs375990626 Chr.1: 74738656-74738656 on Build GRCh38
rs145813190 Chr.1: 74738719-74738719 on Build GRCh38
rs927717809 Chr.1: 74738766-74738766 on Build GRCh38
rs767606379 Chr.1: 74738690-74738690 on Build GRCh38
rs540240280 Chr.1: 74738623-74738623 on Build GRCh38
rs369073129 Chr.1: 74738638-74738638 on Build GRCh38
rs188238254 Chr.1: 74738694-74738694 on Build GRCh38
rs1055553 Chr.1: 74738735-74738735 on Build GRCh38

View Full Product Details