Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48720817-48721042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASIP1
Entrez Gene ID
54922
Gene Name
Ras interacting protein 1
Gene Aliases
RAIN
UniGene
Hs.233955
Gene Chromosome Location
Chr.19: 48720585-48740833 on Build GRCh38
Transcripts
NM_017805.2 XM_011527053.2 XM_011527055.2 XM_017026914.1 XM_017026915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1240015 c.2847G>A p.L949L Chr.19: 48720843-48720843 on Build GRCh38
191601 c.2816G>T p.R939L Chr.19: 48720874-48720874 on Build GRCh38
1712566 c.2779G>A p.D927N Chr.19: 48720911-48720911 on Build GRCh38
6560664 c.2823C>T p.L941L Chr.19: 48720867-48720867 on Build GRCh38
6763310 c.2869C>T p.P957S Chr.19: 48720821-48720821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773610344 Chr.19: 48720966-48720966 on Build GRCh38
rs763285790 Chr.19: 48720967-48720967 on Build GRCh38
rs139276826 Chr.19: 48720948-48720948 on Build GRCh38
rs763816619 Chr.19: 48721012-48721012 on Build GRCh38
rs35799412 Chr.19: 48720918-48720918 on Build GRCh38
rs778979208 Chr.19: 48720945-48720945 on Build GRCh38
rs142687046 Chr.19: 48720850-48720850 on Build GRCh38
rs375759268 Chr.19: 48720994-48720994 on Build GRCh38
rs956672159 Chr.19: 48720997-48720997 on Build GRCh38
rs1031037200 Chr.19: 48720896-48720896 on Build GRCh38
rs368475522 Chr.19: 48720912-48720912 on Build GRCh38
rs773516310 Chr.19: 48720897-48720897 on Build GRCh38
rs368210824 Chr.19: 48721014-48721014 on Build GRCh38
rs752303192 Chr.19: 48720913-48720913 on Build GRCh38
rs1031128178 Chr.19: 48720975-48720975 on Build GRCh38
rs762029834 Chr.19: 48720886-48720886 on Build GRCh38
rs34800326 Chr.19: 48720863-48720863 on Build GRCh38
rs571194489 Chr.19: 48720954-48720954 on Build GRCh38
rs781492022 Chr.19: 48721016-48721017 on Build GRCh38
rs139364919 Chr.19: 48720951-48720951 on Build GRCh38
rs139549287 Chr.19: 48720884-48720884 on Build GRCh38
rs765600142 Chr.19: 48720890-48720890 on Build GRCh38
rs538720938 Chr.19: 48721018-48721018 on Build GRCh38
rs757657219 Chr.19: 48720843-48720844 on Build GRCh38
rs1012388642 Chr.19: 48720923-48720923 on Build GRCh38
rs747036652 Chr.19: 48720877-48720877 on Build GRCh38
rs200381072 Chr.19: 48720845-48720845 on Build GRCh38
rs1018540680 Chr.19: 48720862-48720862 on Build GRCh38
rs771489719 Chr.19: 48720985-48720985 on Build GRCh38
rs201745517 Chr.19: 48720885-48720885 on Build GRCh38
rs751121993 Chr.19: 48720905-48720905 on Build GRCh38
rs766956155 Chr.19: 48720991-48720991 on Build GRCh38
rs778782771 Chr.19: 48721016-48721016 on Build GRCh38
rs770947821 Chr.19: 48720869-48720869 on Build GRCh38
rs748036865 Chr.19: 48720842-48720842 on Build GRCh38
rs140266474 Chr.19: 48720848-48720848 on Build GRCh38
rs768979771 Chr.19: 48720879-48720879 on Build GRCh38
rs775753012 Chr.19: 48720870-48720870 on Build GRCh38
rs993835120 Chr.19: 48720888-48720888 on Build GRCh38
rs760028636 Chr.19: 48720987-48720987 on Build GRCh38
rs751947581 Chr.19: 48720836-48720837 on Build GRCh38
rs989508564 Chr.19: 48721003-48721003 on Build GRCh38
rs763487005 Chr.19: 48720900-48720900 on Build GRCh38
rs1022765772 Chr.19: 48720914-48720914 on Build GRCh38
rs766982541 Chr.19: 48720911-48720911 on Build GRCh38
rs914460028 Chr.19: 48721006-48721006 on Build GRCh38
rs757308590 Chr.19: 48720943-48720943 on Build GRCh38
rs774718182 Chr.19: 48720986-48720986 on Build GRCh38
rs927128636 Chr.19: 48720973-48720973 on Build GRCh38
rs780688660 Chr.19: 48720835-48720835 on Build GRCh38
rs372040193 Chr.19: 48720992-48720992 on Build GRCh38
rs372213059 Chr.19: 48720921-48720921 on Build GRCh38
rs749060514 Chr.19: 48720929-48720929 on Build GRCh38
rs142588540 Chr.19: 48720907-48720907 on Build GRCh38
rs377403982 Chr.19: 48720957-48720957 on Build GRCh38
rs1050036434 Chr.19: 48720839-48720839 on Build GRCh38

View Full Product Details