Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49098359-49098618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QARS
Entrez Gene ID
5859
Gene Name
glutaminyl-tRNA synthetase
Gene Aliases
GLNRS, MSCCA, PRO2195
UniGene
Hs.79322
Gene Chromosome Location
Chr.3: 49095932-49105129 on Build GRCh38
Transcripts
NM_001272073.1 NM_005051.2 NR_073590.1 XM_017006965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6760896 c.1957-9delC p.? Chr.3: 49098489-49098489 on Build GRCh38
5864775 c.2050C>T p.P684S Chr.3: 49098387-49098387 on Build GRCh38
1423871 c.1958G>A p.G653D Chr.3: 49098479-49098479 on Build GRCh38
446689 c.1984G>C p.E662Q Chr.3: 49098453-49098453 on Build GRCh38
7087219 c.2060G>A p.C687Y Chr.3: 49098377-49098377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768876281 Chr.3: 49098563-49098563 on Build GRCh38
rs752320395 Chr.3: 49098450-49098450 on Build GRCh38
rs762780883 Chr.3: 49098409-49098409 on Build GRCh38
rs142441496 Chr.3: 49098404-49098404 on Build GRCh38
rs367835872 Chr.3: 49098490-49098490 on Build GRCh38
rs140225766 Chr.3: 49098446-49098446 on Build GRCh38
rs374312387 Chr.3: 49098435-49098435 on Build GRCh38
rs756380392 Chr.3: 49098419-49098419 on Build GRCh38
rs750619032 Chr.3: 49098391-49098391 on Build GRCh38
rs756075510 Chr.3: 49098516-49098516 on Build GRCh38
rs1037325022 Chr.3: 49098437-49098437 on Build GRCh38
rs954037456 Chr.3: 49098533-49098533 on Build GRCh38
rs865998076 Chr.3: 49098473-49098473 on Build GRCh38
rs748978904 Chr.3: 49098465-49098465 on Build GRCh38
rs878982739 Chr.3: 49098486-49098486 on Build GRCh38
rs532793779 Chr.3: 49098575-49098575 on Build GRCh38
rs367776807 Chr.3: 49098489-49098489 on Build GRCh38
rs1032058964 Chr.3: 49098493-49098493 on Build GRCh38
rs766197862 Chr.3: 49098417-49098417 on Build GRCh38
rs770657741 Chr.3: 49098479-49098479 on Build GRCh38
rs1013287948 Chr.3: 49098480-49098480 on Build GRCh38
rs753102798 Chr.3: 49098500-49098502 on Build GRCh38
rs752555108 Chr.3: 49098515-49098515 on Build GRCh38
rs755672487 Chr.3: 49098458-49098458 on Build GRCh38
rs777447293 Chr.3: 49098520-49098520 on Build GRCh38
rs765497781 Chr.3: 49098379-49098379 on Build GRCh38
rs574420266 Chr.3: 49098502-49098502 on Build GRCh38
rs879055592 Chr.3: 49098483-49098483 on Build GRCh38
rs1034813926 Chr.3: 49098545-49098545 on Build GRCh38
rs905390740 Chr.3: 49098510-49098510 on Build GRCh38
rs976613108 Chr.3: 49098526-49098526 on Build GRCh38
rs13064784 Chr.3: 49098528-49098528 on Build GRCh38
rs747462635 Chr.3: 49098553-49098553 on Build GRCh38
rs753338846 Chr.3: 49098527-49098527 on Build GRCh38
rs747269470 Chr.3: 49098485-49098485 on Build GRCh38
rs371725638 Chr.3: 49098491-49098491 on Build GRCh38
rs1009354427 Chr.3: 49098497-49098497 on Build GRCh38
rs112579679 Chr.3: 49098499-49098499 on Build GRCh38
rs751375938 Chr.3: 49098494-49098494 on Build GRCh38
rs781741003 Chr.3: 49098572-49098572 on Build GRCh38
rs767548783 Chr.3: 49098513-49098513 on Build GRCh38
rs563148564 Chr.3: 49098534-49098534 on Build GRCh38
rs940163832 Chr.3: 49098433-49098433 on Build GRCh38
rs370587517 Chr.3: 49098438-49098438 on Build GRCh38
rs770241224 Chr.3: 49098583-49098583 on Build GRCh38
rs777618796 Chr.3: 49098464-49098464 on Build GRCh38
rs780478501 Chr.3: 49098484-49098484 on Build GRCh38

View Full Product Details