Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 90005082-90005341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF326
Entrez Gene ID
284695
Gene Name
zinc finger protein 326
Gene Aliases
ZAN75, ZIRD, Zfp326, dJ871E2.1
UniGene
Hs.306221
Gene Chromosome Location
Chr.1: 89995095-90035531 on Build GRCh38
Transcripts
NM_001320185.1 NM_181781.3 NM_182975.3 NM_182976.3 XM_005270779.4 XM_011541290.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829369 c.178G>T p.D60Y Chr.1: 90005213-90005213 on Build GRCh38
2259270 c.168A>C p.S56S Chr.1: 90005203-90005203 on Build GRCh38
1194768 c.98G>T p.G33V Chr.1: 90005133-90005133 on Build GRCh38
6064569 c.122G>T p.G41V Chr.1: 90005157-90005157 on Build GRCh38
6829368 c.204T>A p.G68G Chr.1: 90005239-90005239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774229543 Chr.1: 90005150-90005150 on Build GRCh38
rs201147468 Chr.1: 90005213-90005213 on Build GRCh38
rs764318569 Chr.1: 90005210-90005210 on Build GRCh38
rs757731170 Chr.1: 90005226-90005226 on Build GRCh38
rs763239926 Chr.1: 90005201-90005201 on Build GRCh38
rs979400006 Chr.1: 90005142-90005142 on Build GRCh38
rs944248684 Chr.1: 90005242-90005242 on Build GRCh38
rs759755917 Chr.1: 90005168-90005168 on Build GRCh38
rs755568509 Chr.1: 90005266-90005266 on Build GRCh38
rs779635272 Chr.1: 90005129-90005129 on Build GRCh38
rs538103200 Chr.1: 90005180-90005180 on Build GRCh38
rs932852842 Chr.1: 90005199-90005199 on Build GRCh38
rs201849409 Chr.1: 90005233-90005233 on Build GRCh38
rs748855712 Chr.1: 90005274-90005274 on Build GRCh38
rs764705631 Chr.1: 90005288-90005288 on Build GRCh38
rs891281401 Chr.1: 90005229-90005229 on Build GRCh38
rs751099830 Chr.1: 90005234-90005234 on Build GRCh38
rs768079748 Chr.1: 90005238-90005238 on Build GRCh38
rs925283275 Chr.1: 90005193-90005193 on Build GRCh38
rs754403543 Chr.1: 90005301-90005305 on Build GRCh38
rs748047016 Chr.1: 90005152-90005152 on Build GRCh38
rs554264352 Chr.1: 90005296-90005296 on Build GRCh38
rs772129445 Chr.1: 90005297-90005297 on Build GRCh38
rs780658853 Chr.1: 90005108-90005108 on Build GRCh38
rs778546707 Chr.1: 90005294-90005294 on Build GRCh38
rs138323304 Chr.1: 90005163-90005163 on Build GRCh38
rs768273024 Chr.1: 90005136-90005136 on Build GRCh38
rs748985731 Chr.1: 90005134-90005134 on Build GRCh38
rs924256955 Chr.1: 90005120-90005120 on Build GRCh38
rs779475674 Chr.1: 90005108-90005109 on Build GRCh38
rs752006166 Chr.1: 90005220-90005220 on Build GRCh38
rs138516212 Chr.1: 90005284-90005284 on Build GRCh38
rs749926101 Chr.1: 90005259-90005259 on Build GRCh38
rs578150271 Chr.1: 90005270-90005270 on Build GRCh38
rs772180572 Chr.1: 90005153-90005153 on Build GRCh38
rs567627495 Chr.1: 90005109-90005109 on Build GRCh38
rs775515627 Chr.1: 90005194-90005194 on Build GRCh38
rs755762972 Chr.1: 90005119-90005119 on Build GRCh38

View Full Product Details