Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14135768-14135993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
UniGene
Hs.517817
Gene Chromosome Location
Chr.3: 14124940-14143680 on Build GRCh38
Transcripts
NM_024334.2 XM_017007176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6192668 c.882G>A p.E294E Chr.3: 14135908-14135908 on Build GRCh38
1419495 c.882G>T p.E294D Chr.3: 14135908-14135908 on Build GRCh38
5540532 c.821C>T p.P274L Chr.3: 14135847-14135847 on Build GRCh38
6368714 c.842A>G p.D281G Chr.3: 14135868-14135868 on Build GRCh38
5398303 c.865G>A p.G289R Chr.3: 14135891-14135891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780299346 Chr.3: 14135885-14135885 on Build GRCh38
rs202207425 Chr.3: 14135794-14135794 on Build GRCh38
rs139078900 Chr.3: 14135824-14135824 on Build GRCh38
rs769969149 Chr.3: 14135829-14135829 on Build GRCh38
rs761083427 Chr.3: 14135851-14135851 on Build GRCh38
rs762737458 Chr.3: 14135951-14135951 on Build GRCh38
rs730880226 Chr.3: 14135891-14135891 on Build GRCh38
rs771028028 Chr.3: 14135888-14135888 on Build GRCh38
rs978411423 Chr.3: 14135788-14135788 on Build GRCh38
rs267599627 Chr.3: 14135845-14135845 on Build GRCh38
rs780142044 Chr.3: 14135804-14135804 on Build GRCh38
rs377585035 Chr.3: 14135890-14135890 on Build GRCh38
rs779432280 Chr.3: 14135960-14135961 on Build GRCh38
rs774860013 Chr.3: 14135839-14135839 on Build GRCh38
rs778602479 Chr.3: 14135793-14135793 on Build GRCh38
rs747960452 Chr.3: 14135871-14135871 on Build GRCh38
rs201138253 Chr.3: 14135828-14135828 on Build GRCh38
rs766112098 Chr.3: 14135954-14135954 on Build GRCh38
rs532872056 Chr.3: 14135855-14135855 on Build GRCh38
rs757724015 Chr.3: 14135951-14135954 on Build GRCh38
rs139211731 Chr.3: 14135878-14135878 on Build GRCh38
rs1040468507 Chr.3: 14135934-14135934 on Build GRCh38
rs750133976 Chr.3: 14135944-14135944 on Build GRCh38
rs190262194 Chr.3: 14135933-14135933 on Build GRCh38
rs920512368 Chr.3: 14135916-14135916 on Build GRCh38
rs151009158 Chr.3: 14135860-14135860 on Build GRCh38
rs1050368116 Chr.3: 14135930-14135930 on Build GRCh38
rs535423065 Chr.3: 14135837-14135837 on Build GRCh38
rs193922707 Chr.3: 14135823-14135823 on Build GRCh38
rs759993739 Chr.3: 14135840-14135840 on Build GRCh38
rs756294633 Chr.3: 14135870-14135870 on Build GRCh38
rs892585664 Chr.3: 14135956-14135956 on Build GRCh38
rs755018311 Chr.3: 14135866-14135866 on Build GRCh38
rs768035841 Chr.3: 14135848-14135848 on Build GRCh38
rs370443570 Chr.3: 14135925-14135925 on Build GRCh38
rs757974998 Chr.3: 14135802-14135802 on Build GRCh38
rs763026916 Chr.3: 14135836-14135836 on Build GRCh38
rs186530498 Chr.3: 14135879-14135879 on Build GRCh38
rs764861989 Chr.3: 14135854-14135854 on Build GRCh38
rs886039048 Chr.3: 14135887-14135887 on Build GRCh38
rs139842014 Chr.3: 14135822-14135822 on Build GRCh38
rs758007704 Chr.3: 14135857-14135857 on Build GRCh38
rs751086845 Chr.3: 14135863-14135863 on Build GRCh38
rs373729601 Chr.3: 14135932-14135932 on Build GRCh38
rs776056742 Chr.3: 14135929-14135929 on Build GRCh38
rs727504764 Chr.3: 14135876-14135876 on Build GRCh38

View Full Product Details