Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6496030-6496261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB4A
Entrez Gene ID
10382
Gene Name
tubulin beta 4A class IVa
Gene Aliases
DYT4, TUBB4, beta-5
UniGene
Hs.110837
Gene Chromosome Location
Chr.19: 6494319-6502848 on Build GRCh38
Transcripts
NM_001289123.1 NM_001289127.1 NM_001289129.1 NM_001289130.1 NM_001289131.1 NM_006087.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6086192 c.279C>T p.G93G Chr.19: 6496220-6496220 on Build GRCh38
7283000 c.343G>A p.A115T Chr.19: 6496156-6496156 on Build GRCh38
7213336 c.292G>A p.G98S Chr.19: 6496207-6496207 on Build GRCh38
3713081 c.321G>T p.T107T Chr.19: 6496178-6496178 on Build GRCh38
4453628 c.323A>T p.E108V Chr.19: 6496176-6496176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768190406 Chr.19: 6496095-6496095 on Build GRCh38
rs560881201 Chr.19: 6496084-6496084 on Build GRCh38
rs759144466 Chr.19: 6496178-6496178 on Build GRCh38
rs371265470 Chr.19: 6496126-6496126 on Build GRCh38
rs745995021 Chr.19: 6496129-6496129 on Build GRCh38
rs766118356 Chr.19: 6496079-6496079 on Build GRCh38
rs781137307 Chr.19: 6496133-6496133 on Build GRCh38
rs753210565 Chr.19: 6496097-6496097 on Build GRCh38
rs774127285 Chr.19: 6496172-6496172 on Build GRCh38
rs768924966 Chr.19: 6496227-6496236 on Build GRCh38
rs200757174 Chr.19: 6496064-6496064 on Build GRCh38
rs772746260 Chr.19: 6496067-6496067 on Build GRCh38
rs772090741 Chr.19: 6496130-6496130 on Build GRCh38
rs757594393 Chr.19: 6496241-6496241 on Build GRCh38
rs778943389 Chr.19: 6496230-6496231 on Build GRCh38
rs778138515 Chr.19: 6496117-6496117 on Build GRCh38
rs769489513 Chr.19: 6496148-6496148 on Build GRCh38
rs748051542 Chr.19: 6496145-6496145 on Build GRCh38
rs375537381 Chr.19: 6496056-6496056 on Build GRCh38
rs754136059 Chr.19: 6496118-6496118 on Build GRCh38
rs374559176 Chr.19: 6496070-6496070 on Build GRCh38
rs773118700 Chr.19: 6496150-6496150 on Build GRCh38
rs761298089 Chr.19: 6496227-6496227 on Build GRCh38
rs1053260 Chr.19: 6496073-6496073 on Build GRCh38
rs902910254 Chr.19: 6496240-6496240 on Build GRCh38
rs147013122 Chr.19: 6496103-6496103 on Build GRCh38
rs796738621 Chr.19: 6496063-6496063 on Build GRCh38
rs770749775 Chr.19: 6496169-6496169 on Build GRCh38
rs751182929 Chr.19: 6496081-6496081 on Build GRCh38
rs761535706 Chr.19: 6496052-6496052 on Build GRCh38
rs775871196 Chr.19: 6496222-6496222 on Build GRCh38
rs764502394 Chr.19: 6496236-6496236 on Build GRCh38
rs61741669 Chr.19: 6496157-6496157 on Build GRCh38
rs767100230 Chr.19: 6496214-6496214 on Build GRCh38

View Full Product Details