Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68003482-68003723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC93B1
Entrez Gene ID
81622
Gene Name
unc-93 homolog B1 (C. elegans)
Gene Aliases
IIAE1, UNC93, UNC93B, Unc-93B1
UniGene
Hs.502989
Gene Chromosome Location
Chr.11: 67991100-68004125 on Build GRCh38
Transcripts
NM_030930.3 XM_011545291.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638848 c.197C>T p.A66V Chr.11: 68003698-68003698 on Build GRCh38
1605123 c.233A>G p.Y78C Chr.11: 68003662-68003662 on Build GRCh38
4645071 c.178G>A p.V60M Chr.11: 68003717-68003717 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959902696 Chr.11: 68003539-68003539 on Build GRCh38
rs1033921140 Chr.11: 68003535-68003535 on Build GRCh38
rs535977614 Chr.11: 68003605-68003605 on Build GRCh38
rs1009949113 Chr.11: 68003499-68003499 on Build GRCh38
rs763848707 Chr.11: 68003651-68003651 on Build GRCh38
rs954792909 Chr.11: 68003697-68003697 on Build GRCh38
rs985880111 Chr.11: 68003634-68003634 on Build GRCh38
rs752346686 Chr.11: 68003624-68003624 on Build GRCh38
rs568688811 Chr.11: 68003610-68003610 on Build GRCh38
rs1005698959 Chr.11: 68003553-68003553 on Build GRCh38
rs910501727 Chr.11: 68003520-68003520 on Build GRCh38
rs568440748 Chr.11: 68003572-68003572 on Build GRCh38
rs892236391 Chr.11: 68003601-68003601 on Build GRCh38
rs1016576088 Chr.11: 68003660-68003660 on Build GRCh38
rs777293717 Chr.11: 68003609-68003609 on Build GRCh38
rs904729925 Chr.11: 68003556-68003557 on Build GRCh38
rs1022630278 Chr.11: 68003511-68003511 on Build GRCh38
rs993303626 Chr.11: 68003687-68003687 on Build GRCh38
rs370865340 Chr.11: 68003705-68003705 on Build GRCh38
rs376191930 Chr.11: 68003702-68003702 on Build GRCh38
rs1026080172 Chr.11: 68003688-68003688 on Build GRCh38
rs963270596 Chr.11: 68003673-68003673 on Build GRCh38
rs961418578 Chr.11: 68003589-68003589 on Build GRCh38
rs935919731 Chr.11: 68003647-68003647 on Build GRCh38
rs934800040 Chr.11: 68003571-68003571 on Build GRCh38
rs550046931 Chr.11: 68003534-68003534 on Build GRCh38
rs764685964 Chr.11: 68003621-68003621 on Build GRCh38
rs1001631720 Chr.11: 68003529-68003529 on Build GRCh38
rs553988844 Chr.11: 68003635-68003635 on Build GRCh38
rs989446706 Chr.11: 68003506-68003506 on Build GRCh38
rs922704127 Chr.11: 68003611-68003611 on Build GRCh38
rs1045491514 Chr.11: 68003510-68003510 on Build GRCh38
rs906865134 Chr.11: 68003527-68003527 on Build GRCh38
rs1054185641 Chr.11: 68003648-68003648 on Build GRCh38
rs538026807 Chr.11: 68003501-68003501 on Build GRCh38
rs913060101 Chr.11: 68003672-68003672 on Build GRCh38

View Full Product Details