Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20001370-20001627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PP2D1
Entrez Gene ID
151649
Gene Name
protein phosphatase 2C like domain containing 1
Gene Aliases
C3orf48
UniGene
Hs.733161
Gene Chromosome Location
Chr.3: 19979961-20012273 on Build GRCh38
Transcripts
NM_001252657.1 NR_027694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019978 c.641C>T p.S214L Chr.3: 20001479-20001479 on Build GRCh38
3945368 c.570T>A p.T190T Chr.3: 20001550-20001550 on Build GRCh38
7239504 c.553A>G p.M185V Chr.3: 20001567-20001567 on Build GRCh38
5509905 c.612delT p.F204fs*15 Chr.3: 20001508-20001508 on Build GRCh38
4437594 c.706C>A p.P236T Chr.3: 20001414-20001414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs267599758 Chr.3: 20001561-20001561 on Build GRCh38
rs567551896 Chr.3: 20001476-20001476 on Build GRCh38
rs757795617 Chr.3: 20001579-20001579 on Build GRCh38
rs753748406 Chr.3: 20001482-20001482 on Build GRCh38
rs994896147 Chr.3: 20001475-20001475 on Build GRCh38
rs550751104 Chr.3: 20001507-20001508 on Build GRCh38
rs772388599 Chr.3: 20001553-20001553 on Build GRCh38
rs897762871 Chr.3: 20001441-20001441 on Build GRCh38
rs1004338455 Chr.3: 20001477-20001477 on Build GRCh38
rs1036684517 Chr.3: 20001436-20001436 on Build GRCh38
rs965511460 Chr.3: 20001550-20001550 on Build GRCh38
rs566403179 Chr.3: 20001514-20001514 on Build GRCh38
rs1007223267 Chr.3: 20001487-20001487 on Build GRCh38
rs546716751 Chr.3: 20001486-20001486 on Build GRCh38
rs745851976 Chr.3: 20001577-20001577 on Build GRCh38
rs201179163 Chr.3: 20001515-20001515 on Build GRCh38
rs998405385 Chr.3: 20001568-20001568 on Build GRCh38
rs534129147 Chr.3: 20001434-20001434 on Build GRCh38
rs963323396 Chr.3: 20001520-20001520 on Build GRCh38

View Full Product Details