Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34913654-34913927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS2
Entrez Gene ID
55902
Gene Name
acyl-CoA synthetase short-chain family member 2
Gene Aliases
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1
UniGene
Hs.517034
Gene Chromosome Location
Chr.20: 34874942-34927966 on Build GRCh38
Transcripts
NM_001076552.2 NM_001242393.1 NM_018677.3 XM_005260455.1 XM_005260456.1 XM_006723826.1 XM_011528905.1 XM_011528906.1 XM_011528907.1 XM_011528908.1 XM_011528909.2 XM_011528911.1 XM_011528912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792810 c.578G>T p.G193V Chr.20: 34913760-34913760 on Build GRCh38
6592781 c.605G>A p.R202Q Chr.20: 34913787-34913787 on Build GRCh38
6592804 c.493+1G>A p.? Chr.20: 34913826-34913826 on Build GRCh38
4659076 c.643+1G>A p.? Chr.20: 34913826-34913826 on Build GRCh38
4792811 c.578G>T p.G193V Chr.20: 34913760-34913760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758361816 Chr.20: 34913889-34913889 on Build GRCh38
rs772546778 Chr.20: 34913877-34913877 on Build GRCh38
rs752996933 Chr.20: 34913845-34913845 on Build GRCh38
rs139655686 Chr.20: 34913749-34913749 on Build GRCh38
rs890790993 Chr.20: 34913881-34913881 on Build GRCh38
rs752445483 Chr.20: 34913818-34913818 on Build GRCh38
rs769203598 Chr.20: 34913740-34913740 on Build GRCh38
rs771643101 Chr.20: 34913706-34913706 on Build GRCh38
rs185281896 Chr.20: 34913680-34913680 on Build GRCh38
rs774798428 Chr.20: 34913755-34913755 on Build GRCh38
rs758068442 Chr.20: 34913820-34913820 on Build GRCh38
rs916461195 Chr.20: 34913750-34913750 on Build GRCh38
rs765017042 Chr.20: 34913816-34913816 on Build GRCh38
rs143832046 Chr.20: 34913780-34913780 on Build GRCh38
rs756522782 Chr.20: 34913893-34913893 on Build GRCh38
rs371982555 Chr.20: 34913766-34913766 on Build GRCh38
rs979265199 Chr.20: 34913838-34913838 on Build GRCh38
rs753868628 Chr.20: 34913787-34913787 on Build GRCh38
rs754812244 Chr.20: 34913809-34913809 on Build GRCh38
rs780555543 Chr.20: 34913866-34913866 on Build GRCh38
rs373857009 Chr.20: 34913717-34913717 on Build GRCh38
rs941625405 Chr.20: 34913861-34913861 on Build GRCh38
rs867222166 Chr.20: 34913709-34913709 on Build GRCh38
rs756856993 Chr.20: 34913857-34913857 on Build GRCh38
rs746609250 Chr.20: 34913856-34913856 on Build GRCh38
rs8184664 Chr.20: 34913867-34913867 on Build GRCh38
rs375224834 Chr.20: 34913876-34913876 on Build GRCh38
rs368144466 Chr.20: 34913738-34913738 on Build GRCh38
rs746479403 Chr.20: 34913725-34913725 on Build GRCh38
rs769219891 Chr.20: 34913874-34913874 on Build GRCh38
rs765550605 Chr.20: 34913855-34913856 on Build GRCh38
rs565524892 Chr.20: 34913705-34913705 on Build GRCh38
rs763366248 Chr.20: 34913739-34913739 on Build GRCh38
rs918780029 Chr.20: 34913884-34913884 on Build GRCh38
rs766341618 Chr.20: 34913786-34913786 on Build GRCh38
rs750557055 Chr.20: 34913770-34913770 on Build GRCh38
rs986093794 Chr.20: 34913852-34913852 on Build GRCh38
rs777451548 Chr.20: 34913842-34913842 on Build GRCh38
rs770323381 Chr.20: 34913731-34913731 on Build GRCh38

View Full Product Details